Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 160578354-160578615 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LPA
Entrez Gene ID
4018
Gene Name
lipoprotein(a)
Gene Aliases
AK38, APOA, LP
UniGene
Hs.520120
Gene Chromosome Location
Chr.6: 160531483-160666375 on Build GRCh38
Transcripts
NM_005577.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5869714 c.4471+1G>A p.? Chr.6: 160578522-160578522 on Build GRCh38
741286 c.4470A>T p.Q1490H Chr.6: 160578524-160578524 on Build GRCh38
4548161 c.4405G>A p.E1469K Chr.6: 160578589-160578589 on Build GRCh38
5692977 c.4437C>A p.A1479A Chr.6: 160578557-160578557 on Build GRCh38
6702276 c.4461T>C p.P1487P Chr.6: 160578533-160578533 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs547586738 Chr.6: 160578464-160578464 on Build GRCh38
rs768659501 Chr.6: 160578447-160578447 on Build GRCh38
rs780436447 Chr.6: 160578549-160578549 on Build GRCh38
rs898664310 Chr.6: 160578554-160578554 on Build GRCh38
rs891459772 Chr.6: 160578459-160578459 on Build GRCh38
rs773636480 Chr.6: 160578568-160578568 on Build GRCh38
rs545712498 Chr.6: 160578508-160578508 on Build GRCh38
rs541798241 Chr.6: 160578596-160578596 on Build GRCh38
rs200920617 Chr.6: 160578515-160578515 on Build GRCh38
rs763700880 Chr.6: 160578588-160578588 on Build GRCh38
rs904375393 Chr.6: 160578433-160578433 on Build GRCh38
rs996849199 Chr.6: 160578505-160578505 on Build GRCh38
rs377069904 Chr.6: 160578582-160578582 on Build GRCh38
rs939386094 Chr.6: 160578393-160578393 on Build GRCh38
rs759246724 Chr.6: 160578507-160578507 on Build GRCh38
rs369583851 Chr.6: 160578584-160578584 on Build GRCh38
rs751547669 Chr.6: 160578522-160578522 on Build GRCh38
rs755904089 Chr.6: 160578545-160578545 on Build GRCh38
rs189100806 Chr.6: 160578562-160578562 on Build GRCh38
rs917195660 Chr.6: 160578412-160578412 on Build GRCh38
rs1008607815 Chr.6: 160578460-160578460 on Build GRCh38
rs995633013 Chr.6: 160578575-160578575 on Build GRCh38
rs373234964 Chr.6: 160578591-160578591 on Build GRCh38
rs763148795 Chr.6: 160578475-160578484 on Build GRCh38
rs749421119 Chr.6: 160578553-160578553 on Build GRCh38
rs935699216 Chr.6: 160578440-160578440 on Build GRCh38
rs914392880 Chr.6: 160578427-160578427 on Build GRCh38
rs748735983 Chr.6: 160578558-160578558 on Build GRCh38
rs1044209828 Chr.6: 160578430-160578430 on Build GRCh38
rs768559622 Chr.6: 160578478-160578478 on Build GRCh38
rs1052865866 Chr.6: 160578453-160578453 on Build GRCh38
rs948545811 Chr.6: 160578426-160578426 on Build GRCh38
rs200730022 Chr.6: 160578585-160578585 on Build GRCh38
rs781183372 Chr.6: 160578538-160578538 on Build GRCh38
rs900782994 Chr.6: 160578496-160578496 on Build GRCh38
rs757113256 Chr.6: 160578536-160578536 on Build GRCh38
rs992700565 Chr.6: 160578394-160578394 on Build GRCh38
rs762542305 Chr.6: 160578513-160578513 on Build GRCh38
rs558288825 Chr.6: 160578405-160578405 on Build GRCh38
rs750237739 Chr.6: 160578542-160578542 on Build GRCh38
rs932636062 Chr.6: 160578463-160578463 on Build GRCh38
rs979508083 Chr.6: 160578458-160578458 on Build GRCh38
rs749418158 Chr.6: 160578421-160578421 on Build GRCh38
rs774944402 Chr.6: 160578488-160578488 on Build GRCh38
rs577085780 Chr.6: 160578450-160578450 on Build GRCh38
rs773540732 Chr.6: 160578503-160578503 on Build GRCh38
rs947235834 Chr.6: 160578457-160578457 on Build GRCh38
rs768808174 Chr.6: 160578555-160578555 on Build GRCh38
rs988749200 Chr.6: 160578409-160578409 on Build GRCh38
rs759723650 Chr.6: 160578589-160578589 on Build GRCh38

View Full Product Details