Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 13344897-13345122 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEPHS1
Entrez Gene ID
22929
Gene Name
selenophosphate synthetase 1
Gene Aliases
SELD, SPS, SPS1
UniGene
Hs.124027
Gene Chromosome Location
Chr.10: 13317428-13348298 on Build GRCh38
Transcripts
NM_001195602.1 NM_001195604.1 NM_012247.4 XM_017015942.1 XM_017015943.1 XM_017015944.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
458876 c.13G>A p.E5K Chr.10: 13344938-13344938 on Build GRCh38
916314 c.37G>A p.E13K Chr.10: 13344914-13344914 on Build GRCh38
4472709 c.17C>T p.S6F Chr.10: 13344934-13344934 on Build GRCh38
916316 c.8C>T p.T3M Chr.10: 13344943-13344943 on Build GRCh38
109748 c.25C>T p.P9S Chr.10: 13344926-13344926 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1025047261 Chr.10: 13345016-13345016 on Build GRCh38
rs781715713 Chr.10: 13344925-13344925 on Build GRCh38
rs979405519 Chr.10: 13344960-13344960 on Build GRCh38
rs980866609 Chr.10: 13345074-13345074 on Build GRCh38
rs950077800 Chr.10: 13345069-13345069 on Build GRCh38
rs550355392 Chr.10: 13345033-13345033 on Build GRCh38
rs768967807 Chr.10: 13344970-13344970 on Build GRCh38
rs1015217539 Chr.10: 13344964-13344964 on Build GRCh38
rs750927787 Chr.10: 13345000-13345000 on Build GRCh38
rs761731624 Chr.10: 13344992-13344992 on Build GRCh38
rs373966062 Chr.10: 13344952-13344952 on Build GRCh38
rs925240719 Chr.10: 13345079-13345079 on Build GRCh38
rs557774306 Chr.10: 13345013-13345013 on Build GRCh38
rs569650860 Chr.10: 13345057-13345057 on Build GRCh38
rs762907050 Chr.10: 13344956-13344956 on Build GRCh38
rs770232676 Chr.10: 13344951-13344951 on Build GRCh38
rs936688554 Chr.10: 13345083-13345083 on Build GRCh38
rs746381961 Chr.10: 13344936-13344936 on Build GRCh38
rs146099433 Chr.10: 13344926-13344926 on Build GRCh38
rs202197479 Chr.10: 13344978-13344978 on Build GRCh38
rs771273504 Chr.10: 13344924-13344924 on Build GRCh38

View Full Product Details