Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 114790767-114791028 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNFSF15
Entrez Gene ID
9966
Gene Name
tumor necrosis factor superfamily member 15
Gene Aliases
TL1, TL1A, TNLG1B, VEGI, VEGI192A
UniGene
Hs.23349
Gene Chromosome Location
Chr.9: 114784635-114806128 on Build GRCh38
Transcripts
NM_001204344.1 NM_005118.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3982714 c.330T>A p.F110L Chr.9: 114790878-114790878 on Build GRCh38
6805129 c.435A>T p.G145G Chr.9: 114790773-114790773 on Build GRCh38
4812943 c.187C>T p.L63L Chr.9: 114790790-114790790 on Build GRCh38
6485849 c.382A>T p.T128S Chr.9: 114790826-114790826 on Build GRCh38
3926028 c.364G>A p.E122K Chr.9: 114790844-114790844 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1035213331 Chr.9: 114790977-114790977 on Build GRCh38
rs771705541 Chr.9: 114790995-114790996 on Build GRCh38
rs746677666 Chr.9: 114790938-114790938 on Build GRCh38
rs372685008 Chr.9: 114790883-114790883 on Build GRCh38
rs771250916 Chr.9: 114790968-114790968 on Build GRCh38
rs757078623 Chr.9: 114790832-114790832 on Build GRCh38
rs16931748 Chr.9: 114790942-114790942 on Build GRCh38
rs200853967 Chr.9: 114790951-114790951 on Build GRCh38
rs16931745 Chr.9: 114790878-114790878 on Build GRCh38
rs867957370 Chr.9: 114790794-114790794 on Build GRCh38
rs4246905 Chr.9: 114790969-114790969 on Build GRCh38
rs910999466 Chr.9: 114790936-114790936 on Build GRCh38
rs374337566 Chr.9: 114790827-114790827 on Build GRCh38
rs201333233 Chr.9: 114790882-114790882 on Build GRCh38
rs768134246 Chr.9: 114790940-114790940 on Build GRCh38
rs918842271 Chr.9: 114790834-114790834 on Build GRCh38
rs775368530 Chr.9: 114790993-114790993 on Build GRCh38
rs753523598 Chr.9: 114790889-114790889 on Build GRCh38
rs1026134278 Chr.9: 114790911-114790911 on Build GRCh38
rs755215380 Chr.9: 114790791-114790791 on Build GRCh38
rs565550412 Chr.9: 114790891-114790891 on Build GRCh38
rs779892512 Chr.9: 114790862-114790862 on Build GRCh38
rs771882145 Chr.9: 114790858-114790858 on Build GRCh38
rs568808241 Chr.9: 114790833-114790833 on Build GRCh38
rs763957233 Chr.9: 114790888-114790888 on Build GRCh38
rs200046557 Chr.9: 114790818-114790818 on Build GRCh38
rs749946059 Chr.9: 114790805-114790805 on Build GRCh38
rs746621637 Chr.9: 114790867-114790867 on Build GRCh38
rs773380295 Chr.9: 114790872-114790872 on Build GRCh38
rs772203427 Chr.9: 114790985-114790985 on Build GRCh38
rs767557962 Chr.9: 114790884-114790884 on Build GRCh38
rs550636247 Chr.9: 114790898-114790898 on Build GRCh38
rs753019245 Chr.9: 114790885-114790885 on Build GRCh38
rs748489886 Chr.9: 114790922-114790923 on Build GRCh38
rs770252535 Chr.9: 114790870-114790870 on Build GRCh38
rs752880973 Chr.9: 114790817-114790817 on Build GRCh38
rs138319767 Chr.9: 114790970-114790970 on Build GRCh38
rs753883025 Chr.9: 114790831-114790831 on Build GRCh38
rs200936418 Chr.9: 114790896-114790896 on Build GRCh38
rs922298739 Chr.9: 114790966-114790966 on Build GRCh38
rs879653338 Chr.9: 114790869-114790869 on Build GRCh38
rs888774101 Chr.9: 114790810-114790810 on Build GRCh38
rs370931148 Chr.9: 114790816-114790816 on Build GRCh38
rs192783899 Chr.9: 114790809-114790809 on Build GRCh38

View Full Product Details