Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 67891485-67891710 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIRT1
Entrez Gene ID
23411
Gene Name
sirtuin 1
Gene Aliases
SIR2, SIR2L1, SIR2alpha
UniGene
Hs.369779
Gene Chromosome Location
Chr.10: 67884669-67918390 on Build GRCh38
Transcripts
NM_001142498.1 NM_001314049.1 NM_012238.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7425815 c.881A>T p.Q294L Chr.10: 67891493-67891493 on Build GRCh38
4165523 c.874G>T p.D292Y Chr.10: 67891486-67891486 on Build GRCh38
5810481 c.907A>T p.R303* Chr.10: 67891519-67891519 on Build GRCh38
465866 c.877C>T p.P293S Chr.10: 67891489-67891489 on Build GRCh38
3439741 c.942G>A p.K314K Chr.10: 67891554-67891554 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1007754810 Chr.10: 67891601-67891601 on Build GRCh38
rs910596305 Chr.10: 67891656-67891659 on Build GRCh38
rs999045827 Chr.10: 67891638-67891638 on Build GRCh38
rs770324835 Chr.10: 67891592-67891592 on Build GRCh38
rs562845022 Chr.10: 67891587-67891587 on Build GRCh38
rs777591490 Chr.10: 67891627-67891627 on Build GRCh38
rs748596366 Chr.10: 67891590-67891590 on Build GRCh38
rs1031429230 Chr.10: 67891648-67891648 on Build GRCh38
rs746675681 Chr.10: 67891569-67891569 on Build GRCh38
rs959739792 Chr.10: 67891656-67891656 on Build GRCh38
rs200217670 Chr.10: 67891562-67891562 on Build GRCh38
rs200902165 Chr.10: 67891534-67891534 on Build GRCh38
rs780983084 Chr.10: 67891527-67891527 on Build GRCh38
rs976488542 Chr.10: 67891675-67891675 on Build GRCh38
rs780809769 Chr.10: 67891575-67891575 on Build GRCh38
rs1000708655 Chr.10: 67891589-67891589 on Build GRCh38
rs757288862 Chr.10: 67891518-67891518 on Build GRCh38
rs762559903 Chr.10: 67891605-67891605 on Build GRCh38
rs750297088 Chr.10: 67891560-67891560 on Build GRCh38
rs541783768 Chr.10: 67891637-67891637 on Build GRCh38
rs779808715 Chr.10: 67891568-67891568 on Build GRCh38
rs36107781 Chr.10: 67891561-67891561 on Build GRCh38
rs1032150235 Chr.10: 67891644-67891644 on Build GRCh38
rs199885077 Chr.10: 67891596-67891596 on Build GRCh38
rs773542794 Chr.10: 67891595-67891595 on Build GRCh38
rs561753733 Chr.10: 67891674-67891674 on Build GRCh38
rs754385316 Chr.10: 67891570-67891570 on Build GRCh38
rs778823706 Chr.10: 67891536-67891536 on Build GRCh38
rs1021734816 Chr.10: 67891619-67891619 on Build GRCh38

View Full Product Details