Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 152135107-152135366 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYNE1
Entrez Gene ID
23345
Gene Name
spectrin repeat containing nuclear envelope protein 1
Gene Aliases
8B, ARCA1, C6orf98, CPG2, EDMD4, MYNE1, Nesp1, SCAR8, dJ45H2.2
UniGene
Hs.12967
Gene Chromosome Location
Chr.6: 152121684-152637399 on Build GRCh38
Transcripts
NM_033071.3 NM_182961.3 XM_006715407.1 XM_006715408.2 XM_006715409.1 XM_006715410.2 XM_006715411.1 XM_006715412.2 XM_006715413.2 XM_006715414.1 XM_006715415.2 XM_006715416.2 XM_006715417.2 XM_006715420.2 XM_006715421.2 XM_006715422.1 XM_006715423.2 XM_006715424.2 XM_006715425.2 XM_011535641.2 XM_011535642.2 XM_011535643.1 XM_011535644.1 XM_011535645.2 XM_017010608.1 XM_017010609.1 XM_017010610.1 XM_017010611.1 XM_017010612.1 XM_017010613.1 XM_017010614.1 XM_017010615.1 XM_017010616.1 XM_017010617.1 XM_017010618.1 XM_017010619.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4482097 c.25589C>T p.P8530L Chr.6: 152135159-152135159 on Build GRCh38
3723773 c.25612G>T p.E8538* Chr.6: 152135136-152135136 on Build GRCh38
4482094 c.25733C>T p.P8578L Chr.6: 152135159-152135159 on Build GRCh38
7147094 c.24526C>A p.L8176I Chr.6: 152135202-152135202 on Build GRCh38
3621527 c.25732C>T p.P8578S Chr.6: 152135160-152135160 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761524026 Chr.6: 152135159-152135159 on Build GRCh38
rs989761595 Chr.6: 152135210-152135210 on Build GRCh38
rs370566643 Chr.6: 152135156-152135156 on Build GRCh38
rs753899724 Chr.6: 152135226-152135226 on Build GRCh38
rs953970973 Chr.6: 152135173-152135173 on Build GRCh38
rs753005285 Chr.6: 152135275-152135275 on Build GRCh38
rs755039529 Chr.6: 152135255-152135255 on Build GRCh38
rs771414572 Chr.6: 152135142-152135142 on Build GRCh38
rs3818109 Chr.6: 152135340-152135340 on Build GRCh38
rs750227549 Chr.6: 152135177-152135177 on Build GRCh38
rs41291047 Chr.6: 152135141-152135141 on Build GRCh38
rs915641780 Chr.6: 152135289-152135289 on Build GRCh38
rs148417257 Chr.6: 152135270-152135270 on Build GRCh38
rs774875575 Chr.6: 152135146-152135146 on Build GRCh38
rs767484585 Chr.6: 152135165-152135165 on Build GRCh38
rs760698113 Chr.6: 152135186-152135186 on Build GRCh38
rs549500643 Chr.6: 152135314-152135314 on Build GRCh38
rs886043887 Chr.6: 152135196-152135196 on Build GRCh38
rs371504964 Chr.6: 152135240-152135240 on Build GRCh38
rs367714365 Chr.6: 152135258-152135258 on Build GRCh38
rs947189267 Chr.6: 152135294-152135294 on Build GRCh38
rs760833496 Chr.6: 152135320-152135320 on Build GRCh38
rs559811773 Chr.6: 152135188-152135188 on Build GRCh38
rs569292155 Chr.6: 152135325-152135325 on Build GRCh38
rs200178554 Chr.6: 152135157-152135157 on Build GRCh38
rs148104931 Chr.6: 152135328-152135328 on Build GRCh38
rs778082271 Chr.6: 152135130-152135130 on Build GRCh38
rs117389517 Chr.6: 152135303-152135303 on Build GRCh38
rs560682510 Chr.6: 152135302-152135302 on Build GRCh38
rs573608265 Chr.6: 152135203-152135203 on Build GRCh38
rs766268918 Chr.6: 152135190-152135190 on Build GRCh38

View Full Product Details