Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 47413969-47414200 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCAP
Entrez Gene ID
22937
Gene Name
SREBF chaperone
Gene Aliases
-
UniGene
Hs.531789
Gene Chromosome Location
Chr.3: 47413685-47477126 on Build GRCh38
Transcripts
NM_001320044.1 NM_012235.3 XM_005264967.1 XM_005264968.1 XM_005264971.1 XM_011533501.1 XM_011533502.2 XM_017005918.1 XM_017005919.1 XM_017005920.1 XM_017005921.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1328195 c.3652G>A p.G1218S Chr.3: 47414042-47414042 on Build GRCh38
1045599 c.3614G>T p.S1205I Chr.3: 47414080-47414080 on Build GRCh38
7279950 c.3640C>T p.L1214L Chr.3: 47414054-47414054 on Build GRCh38
4795120 c.3677A>G p.D1226G Chr.3: 47414017-47414017 on Build GRCh38
3993114 c.3662G>T p.C1221F Chr.3: 47414032-47414032 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772461126 Chr.3: 47414174-47414174 on Build GRCh38
rs766182543 Chr.3: 47414016-47414016 on Build GRCh38
rs768528532 Chr.3: 47414043-47414043 on Build GRCh38
rs775479060 Chr.3: 47414107-47414107 on Build GRCh38
rs769512736 Chr.3: 47414164-47414165 on Build GRCh38
rs769791731 Chr.3: 47414157-47414157 on Build GRCh38
rs201494816 Chr.3: 47414042-47414042 on Build GRCh38
rs531968147 Chr.3: 47414145-47414145 on Build GRCh38
rs779194978 Chr.3: 47414025-47414025 on Build GRCh38
rs765589857 Chr.3: 47414124-47414124 on Build GRCh38
rs993859488 Chr.3: 47414175-47414175 on Build GRCh38
rs17849133 Chr.3: 47414097-47414097 on Build GRCh38
rs767719888 Chr.3: 47413994-47413994 on Build GRCh38
rs747999475 Chr.3: 47414061-47414061 on Build GRCh38
rs946315837 Chr.3: 47413998-47413998 on Build GRCh38
rs371906022 Chr.3: 47414072-47414072 on Build GRCh38
rs929441875 Chr.3: 47414052-47414052 on Build GRCh38
rs777780298 Chr.3: 47414158-47414158 on Build GRCh38
rs564017528 Chr.3: 47414132-47414132 on Build GRCh38
rs757976509 Chr.3: 47414028-47414028 on Build GRCh38
rs1037523216 Chr.3: 47414092-47414092 on Build GRCh38
rs879130747 Chr.3: 47413990-47413990 on Build GRCh38
rs750780254 Chr.3: 47414128-47414128 on Build GRCh38
rs754193247 Chr.3: 47414015-47414015 on Build GRCh38
rs780909102 Chr.3: 47414160-47414162 on Build GRCh38
rs754056645 Chr.3: 47414059-47414059 on Build GRCh38
rs200561713 Chr.3: 47414007-47414007 on Build GRCh38
rs376828685 Chr.3: 47414133-47414133 on Build GRCh38
rs748133425 Chr.3: 47414150-47414150 on Build GRCh38
rs145894255 Chr.3: 47414034-47414034 on Build GRCh38
rs570212243 Chr.3: 47414000-47414000 on Build GRCh38
rs897636919 Chr.3: 47414113-47414113 on Build GRCh38
rs779735698 Chr.3: 47414141-47414141 on Build GRCh38
rs768331758 Chr.3: 47414139-47414139 on Build GRCh38
rs762114410 Chr.3: 47414102-47414102 on Build GRCh38
rs764639012 Chr.3: 47414013-47414013 on Build GRCh38
rs773084832 Chr.3: 47414074-47414074 on Build GRCh38
rs764036113 Chr.3: 47414118-47414118 on Build GRCh38
rs762248753 Chr.3: 47414120-47414120 on Build GRCh38
rs751071982 Chr.3: 47414142-47414142 on Build GRCh38
rs754547859 Chr.3: 47414143-47414143 on Build GRCh38
rs1044363088 Chr.3: 47414111-47414111 on Build GRCh38
rs922965215 Chr.3: 47414030-47414030 on Build GRCh38
rs137940863 Chr.3: 47414026-47414026 on Build GRCh38
rs1050009439 Chr.3: 47414049-47414049 on Build GRCh38
rs535264774 Chr.3: 47414115-47414115 on Build GRCh38
rs553479595 Chr.3: 47414173-47414173 on Build GRCh38
rs747973512 Chr.3: 47414163-47414164 on Build GRCh38
rs759843483 Chr.3: 47414085-47414085 on Build GRCh38

View Full Product Details