Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 3155540-3155809 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TUBB2A
Entrez Gene ID
7280
Gene Name
tubulin beta 2A class IIa
Gene Aliases
CDCBM5, TUBB, TUBB2
UniGene
Hs.654543
Gene Chromosome Location
Chr.6: 3153666-3157549 on Build GRCh38
Transcripts
NM_001069.2 NM_001310315.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5420226 c.190_191insGA p.I64fs*58 Chr.6: 3155711-3155712 on Build GRCh38
6173654 c.178G>T p.V60L Chr.6: 3155724-3155724 on Build GRCh38
741372 c.205G>A p.E69K Chr.6: 3155697-3155697 on Build GRCh38
7396284 c.249G>T p.Q83H Chr.6: 3155653-3155653 on Build GRCh38
1077598 c.215C>T p.T72I Chr.6: 3155687-3155687 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754390101 Chr.6: 3155584-3155584 on Build GRCh38
rs1133246 Chr.6: 3155635-3155635 on Build GRCh38
rs746335930 Chr.6: 3155646-3155646 on Build GRCh38
rs41300839 Chr.6: 3155713-3155713 on Build GRCh38
rs3209185 Chr.6: 3155680-3155680 on Build GRCh38
rs1054310 Chr.6: 3155662-3155662 on Build GRCh38
rs757935036 Chr.6: 3155691-3155692 on Build GRCh38
rs150597034 Chr.6: 3155626-3155626 on Build GRCh38
rs751381384 Chr.6: 3155590-3155590 on Build GRCh38
rs1007884939 Chr.6: 3155572-3155572 on Build GRCh38
rs747419373 Chr.6: 3155665-3155665 on Build GRCh38
rs769698643 Chr.6: 3155704-3155704 on Build GRCh38
rs542813577 Chr.6: 3155568-3155568 on Build GRCh38
rs759304011 Chr.6: 3155750-3155750 on Build GRCh38
rs767138501 Chr.6: 3155624-3155624 on Build GRCh38
rs562003687 Chr.6: 3155632-3155632 on Build GRCh38
rs762885759 Chr.6: 3155574-3155574 on Build GRCh38
rs995437791 Chr.6: 3155728-3155728 on Build GRCh38
rs752234456 Chr.6: 3155625-3155625 on Build GRCh38
rs76684628 Chr.6: 3155784-3155784 on Build GRCh38
rs11550262 Chr.6: 3155682-3155682 on Build GRCh38
rs376123925 Chr.6: 3155614-3155614 on Build GRCh38
rs1133244 Chr.6: 3155722-3155722 on Build GRCh38
rs3209186 Chr.6: 3155677-3155677 on Build GRCh38
rs754625462 Chr.6: 3155611-3155611 on Build GRCh38
rs766125833 Chr.6: 3155589-3155589 on Build GRCh38
rs752223895 Chr.6: 3155757-3155757 on Build GRCh38
rs755820888 Chr.6: 3155763-3155763 on Build GRCh38
rs753446830 Chr.6: 3155768-3155768 on Build GRCh38
rs774124807 Chr.6: 3155718-3155718 on Build GRCh38
rs1133245 Chr.6: 3155707-3155707 on Build GRCh38
rs142311519 Chr.6: 3155565-3155565 on Build GRCh38
rs763648440 Chr.6: 3155766-3155766 on Build GRCh38
rs758995246 Chr.6: 3155659-3155659 on Build GRCh38
rs776775895 Chr.6: 3155676-3155676 on Build GRCh38
rs758944509 Chr.6: 3155781-3155781 on Build GRCh38
rs1054309 Chr.6: 3155674-3155674 on Build GRCh38
rs1054304 Chr.6: 3155692-3155692 on Build GRCh38
rs115862064 Chr.6: 3155754-3155754 on Build GRCh38
rs768918799 Chr.6: 3155668-3155668 on Build GRCh38
rs1054305 Chr.6: 3155686-3155686 on Build GRCh38

View Full Product Details