Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 84479433-84479694 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPF1
Entrez Gene ID
80135
Gene Name
ribosome production factor 1 homolog
Gene Aliases
BXDC5
UniGene
Hs.481202
Gene Chromosome Location
Chr.1: 84479237-84498350 on Build GRCh38
Transcripts
NM_025065.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5010562 c.214C>T p.Q72* Chr.1: 84479495-84479495 on Build GRCh38
5462764 c.216G>A p.Q72Q Chr.1: 84479497-84479497 on Build GRCh38
211963 c.164C>T p.S55L Chr.1: 84479445-84479445 on Build GRCh38
7415955 c.223C>T p.R75W Chr.1: 84479504-84479504 on Build GRCh38
1474218 c.201C>T p.F67F Chr.1: 84479482-84479482 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs35471136 Chr.1: 84479562-84479562 on Build GRCh38
rs894923073 Chr.1: 84479480-84479480 on Build GRCh38
rs150868852 Chr.1: 84479496-84479496 on Build GRCh38
rs950564780 Chr.1: 84479664-84479664 on Build GRCh38
rs746522688 Chr.1: 84479470-84479470 on Build GRCh38
rs758264553 Chr.1: 84479484-84479484 on Build GRCh38
rs762577656 Chr.1: 84479536-84479536 on Build GRCh38
rs770236342 Chr.1: 84479479-84479479 on Build GRCh38
rs776507178 Chr.1: 84479519-84479519 on Build GRCh38
rs1040063467 Chr.1: 84479608-84479608 on Build GRCh38
rs768920556 Chr.1: 84479491-84479491 on Build GRCh38
rs746238125 Chr.1: 84479478-84479478 on Build GRCh38
rs754103650 Chr.1: 84479557-84479563 on Build GRCh38
rs764481130 Chr.1: 84479551-84479551 on Build GRCh38
rs17374048 Chr.1: 84479656-84479656 on Build GRCh38
rs749564354 Chr.1: 84479487-84479487 on Build GRCh38
rs964780934 Chr.1: 84479628-84479628 on Build GRCh38
rs987462074 Chr.1: 84479583-84479583 on Build GRCh38
rs988525592 Chr.1: 84479576-84479576 on Build GRCh38
rs754453855 Chr.1: 84479547-84479547 on Build GRCh38
rs780748679 Chr.1: 84479649-84479649 on Build GRCh38
rs199963998 Chr.1: 84479517-84479517 on Build GRCh38
rs763786742 Chr.1: 84479542-84479542 on Build GRCh38
rs887727725 Chr.1: 84479663-84479663 on Build GRCh38
rs760608096 Chr.1: 84479523-84479523 on Build GRCh38
rs781770314 Chr.1: 84479557-84479557 on Build GRCh38
rs375909828 Chr.1: 84479477-84479477 on Build GRCh38
rs983297955 Chr.1: 84479670-84479670 on Build GRCh38
rs757760874 Chr.1: 84479475-84479475 on Build GRCh38
rs752455543 Chr.1: 84479533-84479533 on Build GRCh38
rs545982996 Chr.1: 84479485-84479485 on Build GRCh38
rs369482297 Chr.1: 84479552-84479552 on Build GRCh38
rs546351541 Chr.1: 84479604-84479604 on Build GRCh38
rs1049349220 Chr.1: 84479661-84479661 on Build GRCh38
rs751043500 Chr.1: 84479545-84479545 on Build GRCh38
rs765062316 Chr.1: 84479532-84479532 on Build GRCh38
rs543914256 Chr.1: 84479463-84479463 on Build GRCh38
rs779997935 Chr.1: 84479492-84479492 on Build GRCh38
rs761897072 Chr.1: 84479516-84479516 on Build GRCh38
rs759343704 Chr.1: 84479525-84479525 on Build GRCh38
rs756659309 Chr.1: 84479559-84479559 on Build GRCh38
rs751017716 Chr.1: 84479558-84479558 on Build GRCh38
rs912872597 Chr.1: 84479577-84479577 on Build GRCh38
rs34550907 Chr.1: 84479534-84479534 on Build GRCh38

View Full Product Details