Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4740381-4740654 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZMYND15
Entrez Gene ID
84225
Gene Name
zinc finger MYND-type containing 15
Gene Aliases
SPGF14
UniGene
Hs.47223
Gene Chromosome Location
Chr.17: 4740015-4746119 on Build GRCh38
Transcripts
NM_001136046.2 NM_001267822.1 NM_032265.2 XM_017025218.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6827645 c.68G>A p.R23Q Chr.17: 4740616-4740616 on Build GRCh38
3518674 c.33C>T p.F11F Chr.17: 4740581-4740581 on Build GRCh38
1172430 c.95T>C p.V32A Chr.17: 4740643-4740643 on Build GRCh38
1172431 c.95T>C p.V32A Chr.17: 4740643-4740643 on Build GRCh38
3518673 c.33C>T p.F11F Chr.17: 4740581-4740581 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774347238 Chr.17: 4740539-4740539 on Build GRCh38
rs758319202 Chr.17: 4740597-4740597 on Build GRCh38
rs763465783 Chr.17: 4740528-4740528 on Build GRCh38
rs533117259 Chr.17: 4740478-4740478 on Build GRCh38
rs767713083 Chr.17: 4740548-4740548 on Build GRCh38
rs779665279 Chr.17: 4740509-4740509 on Build GRCh38
rs200325434 Chr.17: 4740518-4740518 on Build GRCh38
rs748591091 Chr.17: 4740627-4740627 on Build GRCh38
rs766398795 Chr.17: 4740533-4740533 on Build GRCh38
rs780957533 Chr.17: 4740614-4740614 on Build GRCh38
rs906311678 Chr.17: 4740467-4740467 on Build GRCh38
rs186245265 Chr.17: 4740406-4740406 on Build GRCh38
rs375316470 Chr.17: 4740505-4740505 on Build GRCh38
rs11655692 Chr.17: 4740499-4740499 on Build GRCh38
rs759766657 Chr.17: 4740544-4740544 on Build GRCh38
rs144585230 Chr.17: 4740616-4740616 on Build GRCh38
rs757826463 Chr.17: 4740508-4740508 on Build GRCh38
rs751987797 Chr.17: 4740606-4740606 on Build GRCh38
rs369871047 Chr.17: 4740527-4740527 on Build GRCh38
rs748453021 Chr.17: 4740525-4740525 on Build GRCh38
rs1027671060 Chr.17: 4740529-4740529 on Build GRCh38
rs989985605 Chr.17: 4740404-4740404 on Build GRCh38
rs779718566 Chr.17: 4740605-4740605 on Build GRCh38
rs750351512 Chr.17: 4740571-4740571 on Build GRCh38
rs754739451 Chr.17: 4740607-4740607 on Build GRCh38
rs549281875 Chr.17: 4740434-4740434 on Build GRCh38
rs753700368 Chr.17: 4740550-4740550 on Build GRCh38
rs940306972 Chr.17: 4740480-4740480 on Build GRCh38
rs1036034220 Chr.17: 4740497-4740497 on Build GRCh38
rs765269596 Chr.17: 4740565-4740565 on Build GRCh38
rs766786692 Chr.17: 4740526-4740526 on Build GRCh38
rs773583128 Chr.17: 4740633-4740633 on Build GRCh38
rs947905459 Chr.17: 4740414-4740414 on Build GRCh38
rs746693705 Chr.17: 4740516-4740516 on Build GRCh38
rs775725297 Chr.17: 4740548-4740549 on Build GRCh38
rs770073986 Chr.17: 4740631-4740631 on Build GRCh38
rs146442451 Chr.17: 4740458-4740458 on Build GRCh38
rs865993897 Chr.17: 4740415-4740415 on Build GRCh38
rs777152925 Chr.17: 4740524-4740524 on Build GRCh38
rs756972315 Chr.17: 4740562-4740562 on Build GRCh38

View Full Product Details