Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 102195063-102195288 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR20
Entrez Gene ID
91833
Gene Name
WD repeat domain 20
Gene Aliases
DMR
UniGene
Hs.36859
Gene Chromosome Location
Chr.14: 102139518-102224686 on Build GRCh38
Transcripts
NM_001242414.1 NM_001242415.1 NM_001242416.1 NM_001242417.1 NM_001242418.1 NM_001320130.1 NM_144574.3 NM_181291.2 NM_181308.2 NR_038354.1 XM_006720308.1 XM_006720309.1 XM_006720310.1 XM_006720311.1 XM_006720313.2 XM_006720318.2 XM_006720320.3 XM_011537335.2 XM_011537336.2 XM_011537337.2 XM_011537338.1 XM_011537339.2 XM_011537340.2 XM_011537341.2 XM_011537343.1 XM_011537344.2 XM_011537345.1 XM_011537346.1 XM_011537347.1 XM_011537352.2 XM_011537353.2 XM_011537355.2 XM_011537356.2 XM_011537358.2 XM_017021766.1 XM_017021767.1 XM_017021768.1 XM_017021770.1 XM_017021771.1 XM_017021772.1 XM_017021773.1 XM_017021774.1 XM_017021775.1 XM_017021776.1 XM_017021777.1 XM_017021778.1 XM_017021779.1 XM_017021780.1 XM_017021781.1 XM_017021782.1 XM_017021783.1 XM_017021784.1 XM_017021785.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6340840 c.389T>C p.I130T Chr.14: 102195077-102195077 on Build GRCh38
7140504 c.399C>A p.I133I Chr.14: 102195087-102195087 on Build GRCh38
4493305 c.376C>T p.Q126* Chr.14: 102195064-102195064 on Build GRCh38
6821622 c.415A>T p.K139* Chr.14: 102195103-102195103 on Build GRCh38
6821623 c.415A>T p.K139* Chr.14: 102195103-102195103 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754996612 Chr.14: 102195235-102195236 on Build GRCh38
rs370321551 Chr.14: 102195133-102195134 on Build GRCh38
rs770519641 Chr.14: 102195153-102195153 on Build GRCh38
rs893970843 Chr.14: 102195212-102195212 on Build GRCh38
rs769010773 Chr.14: 102195100-102195100 on Build GRCh38
rs575972373 Chr.14: 102195254-102195254 on Build GRCh38
rs775975313 Chr.14: 102195174-102195174 on Build GRCh38
rs908608417 Chr.14: 102195191-102195191 on Build GRCh38
rs772756005 Chr.14: 102195132-102195132 on Build GRCh38
rs749640292 Chr.14: 102195098-102195098 on Build GRCh38
rs114354673 Chr.14: 102195213-102195213 on Build GRCh38
rs955576609 Chr.14: 102195156-102195156 on Build GRCh38
rs1004768525 Chr.14: 102195237-102195237 on Build GRCh38
rs920091084 Chr.14: 102195216-102195216 on Build GRCh38
rs938208752 Chr.14: 102195229-102195229 on Build GRCh38
rs541767714 Chr.14: 102195258-102195258 on Build GRCh38
rs894117721 Chr.14: 102195238-102195238 on Build GRCh38
rs78964001 Chr.14: 102195089-102195089 on Build GRCh38
rs1055395284 Chr.14: 102195235-102195235 on Build GRCh38
rs760200190 Chr.14: 102195140-102195140 on Build GRCh38
rs772551709 Chr.14: 102195117-102195117 on Build GRCh38
rs377478552 Chr.14: 102195205-102195205 on Build GRCh38
rs1052645502 Chr.14: 102195210-102195210 on Build GRCh38
rs147480784 Chr.14: 102195236-102195236 on Build GRCh38
rs940157160 Chr.14: 102195207-102195207 on Build GRCh38
rs779393048 Chr.14: 102195104-102195104 on Build GRCh38
rs776297826 Chr.14: 102195167-102195167 on Build GRCh38
rs756333246 Chr.14: 102195088-102195088 on Build GRCh38
rs975309583 Chr.14: 102195144-102195144 on Build GRCh38
rs759263130 Chr.14: 102195170-102195170 on Build GRCh38

View Full Product Details