Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 234319959-234320230 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC35F3
Entrez Gene ID
148641
Gene Name
solute carrier family 35 member F3
Gene Aliases
-
UniGene
Hs.158748
Gene Chromosome Location
Chr.1: 233904712-234324518 on Build GRCh38
Transcripts
NM_001300845.1 NM_173508.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5799109 c.1030+9A>G p.? Chr.1: 234320196-234320196 on Build GRCh38
213663 c.1213G>A p.V405I Chr.1: 234320163-234320163 on Build GRCh38
6125307 c.1027G>T p.A343S Chr.1: 234320184-234320184 on Build GRCh38
6125306 c.1234G>T p.A412S Chr.1: 234320184-234320184 on Build GRCh38
7183615 c.1197G>C p.L399L Chr.1: 234320147-234320147 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1001853207 Chr.1: 234320148-234320148 on Build GRCh38
rs761489653 Chr.1: 234320075-234320077 on Build GRCh38
rs778380324 Chr.1: 234320103-234320103 on Build GRCh38
rs75778483 Chr.1: 234320014-234320014 on Build GRCh38
rs771529879 Chr.1: 234320109-234320109 on Build GRCh38
rs370569228 Chr.1: 234320074-234320074 on Build GRCh38
rs371861159 Chr.1: 234320129-234320129 on Build GRCh38
rs746880050 Chr.1: 234320049-234320049 on Build GRCh38
rs35690609 Chr.1: 234320038-234320039 on Build GRCh38
rs921169852 Chr.1: 234319986-234319986 on Build GRCh38
rs888024731 Chr.1: 234320135-234320135 on Build GRCh38
rs542554084 Chr.1: 234319981-234319981 on Build GRCh38
rs149597390 Chr.1: 234320163-234320163 on Build GRCh38
rs762786015 Chr.1: 234320199-234320199 on Build GRCh38
rs539243199 Chr.1: 234320107-234320107 on Build GRCh38
rs377116741 Chr.1: 234320130-234320130 on Build GRCh38
rs148813050 Chr.1: 234320075-234320075 on Build GRCh38
rs1046144348 Chr.1: 234320110-234320110 on Build GRCh38
rs776288448 Chr.1: 234320140-234320140 on Build GRCh38
rs755148816 Chr.1: 234320066-234320066 on Build GRCh38
rs931237988 Chr.1: 234319990-234319990 on Build GRCh38
rs374042969 Chr.1: 234320200-234320200 on Build GRCh38
rs748214286 Chr.1: 234320087-234320087 on Build GRCh38
rs770014497 Chr.1: 234320101-234320101 on Build GRCh38
rs945743449 Chr.1: 234319995-234319995 on Build GRCh38
rs767812446 Chr.1: 234319982-234319982 on Build GRCh38
rs139635407 Chr.1: 234320180-234320180 on Build GRCh38
rs761274959 Chr.1: 234320162-234320162 on Build GRCh38

View Full Product Details