Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76152266-76152499 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF157
Entrez Gene ID
114804
Gene Name
ring finger protein 157
Gene Aliases
-
UniGene
Hs.500643
Gene Chromosome Location
Chr.17: 76142453-76240368 on Build GRCh38
Transcripts
NM_052916.2 XM_005257007.4 XM_017024117.1 XM_017024118.1 XM_017024119.1 XM_017024120.1 NR_040017.1

Gene Symbol
RNF157-AS1
Entrez Gene ID
100507218
Gene Name
RNF157 antisense RNA 1
Gene Aliases
-
UniGene
Hs.654866
Gene Chromosome Location
Chr.17: 76140556-76154648 on Build GRCh38
Transcripts
NM_052916.2 XM_005257007.4 XM_017024117.1 XM_017024118.1 XM_017024119.1 XM_017024120.1 NR_040017.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4862028 c.1864G>C p.D622H Chr.17: 76152412-76152412 on Build GRCh38
4786740 c.1820C>T p.T607M Chr.17: 76152456-76152456 on Build GRCh38
5907703 c.1904C>T p.S635F Chr.17: 76152372-76152372 on Build GRCh38
5657745 c.1873A>C p.S625R Chr.17: 76152403-76152403 on Build GRCh38
7097603 c.1869C>T p.I623I Chr.17: 76152407-76152407 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs115220134 Chr.17: 76152455-76152455 on Build GRCh38
rs149285969 Chr.17: 76152400-76152400 on Build GRCh38
rs199996462 Chr.17: 76152467-76152467 on Build GRCh38
rs771325661 Chr.17: 76152325-76152325 on Build GRCh38
rs143725203 Chr.17: 76152452-76152452 on Build GRCh38
rs924029254 Chr.17: 76152287-76152287 on Build GRCh38
rs887214444 Chr.17: 76152371-76152371 on Build GRCh38
rs771855433 Chr.17: 76152460-76152460 on Build GRCh38
rs537147365 Chr.17: 76152451-76152451 on Build GRCh38
rs148503232 Chr.17: 76152456-76152456 on Build GRCh38
rs773917769 Chr.17: 76152340-76152340 on Build GRCh38
rs746575050 Chr.17: 76152464-76152464 on Build GRCh38
rs61760885 Chr.17: 76152363-76152363 on Build GRCh38
rs779796224 Chr.17: 76152461-76152461 on Build GRCh38
rs915378335 Chr.17: 76152387-76152387 on Build GRCh38
rs544995207 Chr.17: 76152468-76152468 on Build GRCh38
rs140091781 Chr.17: 76152406-76152406 on Build GRCh38
rs768491970 Chr.17: 76152393-76152393 on Build GRCh38
rs922835877 Chr.17: 76152432-76152432 on Build GRCh38
rs910236477 Chr.17: 76152291-76152291 on Build GRCh38
rs935394765 Chr.17: 76152341-76152341 on Build GRCh38
rs1039983915 Chr.17: 76152299-76152299 on Build GRCh38
rs769565682 Chr.17: 76152473-76152473 on Build GRCh38
rs767684454 Chr.17: 76152416-76152416 on Build GRCh38
rs894490254 Chr.17: 76152408-76152408 on Build GRCh38
rs765048782 Chr.17: 76152402-76152402 on Build GRCh38
rs778215506 Chr.17: 76152446-76152446 on Build GRCh38
rs773019051 Chr.17: 76152476-76152476 on Build GRCh38
rs901423484 Chr.17: 76152333-76152333 on Build GRCh38
rs143071812 Chr.17: 76152348-76152348 on Build GRCh38
rs759772932 Chr.17: 76152407-76152407 on Build GRCh38
rs932992175 Chr.17: 76152444-76152444 on Build GRCh38
rs761786557 Chr.17: 76152472-76152472 on Build GRCh38
rs761580440 Chr.17: 76152401-76152401 on Build GRCh38
rs752774588 Chr.17: 76152430-76152430 on Build GRCh38
rs771802307 Chr.17: 76152359-76152359 on Build GRCh38

View Full Product Details