Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 44784610-44784857 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BEST4
Entrez Gene ID
266675
Gene Name
bestrophin 4
Gene Aliases
VMD2L2
UniGene
Hs.302513
Gene Chromosome Location
Chr.1: 44781836-44792828 on Build GRCh38
Transcripts
NM_153274.2 XM_017001020.1 XM_017001021.1 XM_017001022.1 XM_017001023.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4459278 c.1119C>T p.P373P Chr.1: 44784658-44784658 on Build GRCh38
7425897 c.1104C>A p.A368A Chr.1: 44784673-44784673 on Build GRCh38
4439776 c.1032T>C p.L344L Chr.1: 44784745-44784745 on Build GRCh38
4008436 c.1037C>T p.P346L Chr.1: 44784740-44784740 on Build GRCh38
6611369 c.1009G>A p.V337M Chr.1: 44784768-44784768 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs966114563 Chr.1: 44784647-44784647 on Build GRCh38
rs777990349 Chr.1: 44784674-44784674 on Build GRCh38
rs768444892 Chr.1: 44784819-44784819 on Build GRCh38
rs776266325 Chr.1: 44784702-44784702 on Build GRCh38
rs756572035 Chr.1: 44784673-44784673 on Build GRCh38
rs753116814 Chr.1: 44784710-44784715 on Build GRCh38
rs750247002 Chr.1: 44784787-44784787 on Build GRCh38
rs755503437 Chr.1: 44784664-44784664 on Build GRCh38
rs779163006 Chr.1: 44784682-44784682 on Build GRCh38
rs762584742 Chr.1: 44784832-44784832 on Build GRCh38
rs779216302 Chr.1: 44784772-44784772 on Build GRCh38
rs767954927 Chr.1: 44784669-44784669 on Build GRCh38
rs546689077 Chr.1: 44784724-44784724 on Build GRCh38
rs770637220 Chr.1: 44784816-44784816 on Build GRCh38
rs761565620 Chr.1: 44784703-44784703 on Build GRCh38
rs372933274 Chr.1: 44784823-44784823 on Build GRCh38
rs761357873 Chr.1: 44784634-44784634 on Build GRCh38
rs745942679 Chr.1: 44784780-44784780 on Build GRCh38
rs745891320 Chr.1: 44784685-44784685 on Build GRCh38
rs1039535996 Chr.1: 44784721-44784721 on Build GRCh38
rs758614240 Chr.1: 44784781-44784781 on Build GRCh38
rs772723521 Chr.1: 44784710-44784710 on Build GRCh38
rs749575880 Chr.1: 44784679-44784679 on Build GRCh38
rs988031054 Chr.1: 44784743-44784743 on Build GRCh38
rs776518836 Chr.1: 44784821-44784821 on Build GRCh38
rs942555009 Chr.1: 44784716-44784716 on Build GRCh38
rs138819993 Chr.1: 44784739-44784739 on Build GRCh38
rs533175337 Chr.1: 44784696-44784696 on Build GRCh38
rs754115252 Chr.1: 44784746-44784746 on Build GRCh38
rs772145581 Chr.1: 44784695-44784695 on Build GRCh38
rs772916692 Chr.1: 44784831-44784831 on Build GRCh38
rs997168788 Chr.1: 44784671-44784671 on Build GRCh38
rs765870318 Chr.1: 44784720-44784720 on Build GRCh38
rs566574541 Chr.1: 44784807-44784807 on Build GRCh38
rs1029183229 Chr.1: 44784678-44784678 on Build GRCh38
rs756736992 Chr.1: 44784810-44784811 on Build GRCh38
rs779983381 Chr.1: 44784799-44784799 on Build GRCh38
rs764563258 Chr.1: 44784742-44784742 on Build GRCh38
rs762383507 Chr.1: 44784715-44784715 on Build GRCh38
rs1041631425 Chr.1: 44784783-44784783 on Build GRCh38
rs917816812 Chr.1: 44784786-44784786 on Build GRCh38
rs749901864 Chr.1: 44784656-44784656 on Build GRCh38
rs141308464 Chr.1: 44784704-44784704 on Build GRCh38
rs564255785 Chr.1: 44784680-44784680 on Build GRCh38
rs768552808 Chr.1: 44784698-44784698 on Build GRCh38
rs757574175 Chr.1: 44784747-44784747 on Build GRCh38
rs1002510939 Chr.1: 44784812-44784812 on Build GRCh38
rs925120042 Chr.1: 44784706-44784706 on Build GRCh38
rs765967549 Chr.1: 44784835-44784835 on Build GRCh38
rs769443238 Chr.1: 44784826-44784826 on Build GRCh38
rs753008617 Chr.1: 44784672-44784672 on Build GRCh38
rs376989433 Chr.1: 44784652-44784652 on Build GRCh38

View Full Product Details