Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 130164281-130164538 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMPD4
Entrez Gene ID
55627
Gene Name
sphingomyelin phosphodiesterase 4
Gene Aliases
NET13, NSMASE-3, NSMASE3
UniGene
Hs.516450
Gene Chromosome Location
Chr.2: 130151392-130182750 on Build GRCh38
Transcripts
NM_001171083.2 NM_017751.4 NM_017951.4 NR_033230.2 NR_033231.2 NR_033232.2 XM_011511443.2 XM_011511444.2 XM_011511445.2 XM_017004448.1 XM_017004449.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3797960 c.919G>A p.E307K Chr.2: 130164436-130164436 on Build GRCh38
5058055 c.954T>C p.Y318Y Chr.2: 130164401-130164401 on Build GRCh38
1399361 c.961delA p.M321fs*53 Chr.2: 130164394-130164394 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs772145567 Chr.2: 130164374-130164374 on Build GRCh38
rs572089027 Chr.2: 130164491-130164491 on Build GRCh38
rs775497956 Chr.2: 130164381-130164381 on Build GRCh38
rs371358700 Chr.2: 130164332-130164332 on Build GRCh38
rs746990306 Chr.2: 130164494-130164494 on Build GRCh38
rs760575086 Chr.2: 130164382-130164382 on Build GRCh38
rs781462260 Chr.2: 130164432-130164432 on Build GRCh38
rs750526844 Chr.2: 130164405-130164405 on Build GRCh38
rs377556845 Chr.2: 130164356-130164356 on Build GRCh38
rs1032377762 Chr.2: 130164302-130164302 on Build GRCh38
rs775589601 Chr.2: 130164493-130164493 on Build GRCh38
rs973571415 Chr.2: 130164510-130164510 on Build GRCh38
rs570987129 Chr.2: 130164305-130164305 on Build GRCh38
rs956928791 Chr.2: 130164313-130164315 on Build GRCh38
rs145962680 Chr.2: 130164398-130164398 on Build GRCh38
rs367702545 Chr.2: 130164463-130164463 on Build GRCh38
rs747483164 Chr.2: 130164333-130164333 on Build GRCh38
rs750484822 Chr.2: 130164327-130164327 on Build GRCh38
rs199586464 Chr.2: 130164365-130164365 on Build GRCh38
rs777436075 Chr.2: 130164344-130164344 on Build GRCh38
rs755360957 Chr.2: 130164431-130164431 on Build GRCh38
rs373373382 Chr.2: 130164342-130164342 on Build GRCh38
rs941826195 Chr.2: 130164505-130164505 on Build GRCh38
rs372842719 Chr.2: 130164326-130164326 on Build GRCh38
rs369981973 Chr.2: 130164316-130164316 on Build GRCh38
rs766878987 Chr.2: 130164422-130164422 on Build GRCh38
rs778567594 Chr.2: 130164475-130164475 on Build GRCh38
rs776464452 Chr.2: 130164384-130164384 on Build GRCh38
rs766939459 Chr.2: 130164516-130164517 on Build GRCh38
rs758894527 Chr.2: 130164330-130164330 on Build GRCh38
rs374746833 Chr.2: 130164371-130164371 on Build GRCh38
rs376121398 Chr.2: 130164331-130164331 on Build GRCh38
rs770599589 Chr.2: 130164361-130164361 on Build GRCh38
rs758416623 Chr.2: 130164417-130164417 on Build GRCh38
rs748519324 Chr.2: 130164446-130164446 on Build GRCh38
rs370508621 Chr.2: 130164487-130164487 on Build GRCh38
rs375408836 Chr.2: 130164496-130164496 on Build GRCh38
rs761981300 Chr.2: 130164387-130164387 on Build GRCh38

View Full Product Details