Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 103573028-103573269 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMY2B
Entrez Gene ID
280
Gene Name
amylase, alpha 2B (pancreatic)
Gene Aliases
AMY2, AMY3, HXA
UniGene
Hs.484588
Gene Chromosome Location
Chr.1: 103554644-103579534 on Build GRCh38
Transcripts
NM_020978.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5080965 c.388T>A p.C130S Chr.1: 103573135-103573135 on Build GRCh38
7091557 c.492G>T p.E164D Chr.1: 103573239-103573239 on Build GRCh38
6264014 c.353C>G p.S118C Chr.1: 103573100-103573100 on Build GRCh38
3470544 c.407C>T p.P136L Chr.1: 103573154-103573154 on Build GRCh38
1244918 c.465A>T p.K155N Chr.1: 103573212-103573212 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs564253866 Chr.1: 103573062-103573062 on Build GRCh38
rs746781812 Chr.1: 103573093-103573093 on Build GRCh38
rs766113372 Chr.1: 103573157-103573157 on Build GRCh38
rs112279755 Chr.1: 103573082-103573082 on Build GRCh38
rs561651261 Chr.1: 103573237-103573237 on Build GRCh38
rs763147263 Chr.1: 103573098-103573098 on Build GRCh38
rs753454981 Chr.1: 103573072-103573072 on Build GRCh38
rs779657016 Chr.1: 103573069-103573069 on Build GRCh38
rs758463862 Chr.1: 103573171-103573171 on Build GRCh38
rs753376439 Chr.1: 103573161-103573161 on Build GRCh38
rs528267080 Chr.1: 103573081-103573081 on Build GRCh38
rs752704778 Chr.1: 103573170-103573170 on Build GRCh38
rs780335193 Chr.1: 103573201-103573201 on Build GRCh38
rs1036967333 Chr.1: 103573123-103573123 on Build GRCh38
rs199958657 Chr.1: 103573206-103573206 on Build GRCh38
rs775566780 Chr.1: 103573094-103573094 on Build GRCh38
rs543472077 Chr.1: 103573204-103573204 on Build GRCh38
rs751994697 Chr.1: 103573055-103573055 on Build GRCh38
rs774923793 Chr.1: 103573107-103573107 on Build GRCh38
rs778250286 Chr.1: 103573164-103573164 on Build GRCh38
rs139026811 Chr.1: 103573084-103573084 on Build GRCh38
rs768701978 Chr.1: 103573194-103573194 on Build GRCh38
rs761042025 Chr.1: 103573230-103573230 on Build GRCh38
rs766587639 Chr.1: 103573232-103573232 on Build GRCh38
rs774244667 Chr.1: 103573207-103573207 on Build GRCh38
rs755109874 Chr.1: 103573067-103573067 on Build GRCh38
rs776379141 Chr.1: 103573236-103573236 on Build GRCh38
rs777825478 Chr.1: 103573172-103573172 on Build GRCh38
rs750106406 Chr.1: 103573066-103573066 on Build GRCh38
rs777664095 Chr.1: 103573092-103573092 on Build GRCh38
rs748203361 Chr.1: 103573213-103573213 on Build GRCh38
rs762266801 Chr.1: 103573108-103573108 on Build GRCh38
rs4098273 Chr.1: 103573100-103573100 on Build GRCh38
rs778925456 Chr.1: 103573083-103573083 on Build GRCh38
rs761023832 Chr.1: 103573155-103573155 on Build GRCh38
rs746863486 Chr.1: 103573176-103573176 on Build GRCh38
rs772607021 Chr.1: 103573220-103573220 on Build GRCh38
rs773696201 Chr.1: 103573221-103573221 on Build GRCh38
rs767889357 Chr.1: 103573109-103573109 on Build GRCh38
rs750835242 Chr.1: 103573122-103573122 on Build GRCh38
rs770833106 Chr.1: 103573200-103573200 on Build GRCh38

View Full Product Details