Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43255720-43255993 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTBK1
Entrez Gene ID
84630
Gene Name
tau tubulin kinase 1
Gene Aliases
BDTK
UniGene
Hs.485436
Gene Chromosome Location
Chr.6: 43243484-43288259 on Build GRCh38
Transcripts
NM_032538.1 XM_011514947.2 XM_017011364.1 XM_017011365.1 XM_017011366.1 XM_017011367.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1664738 c.756G>C p.K252N Chr.6: 43255751-43255751 on Build GRCh38
7127949 c.795G>T p.P265P Chr.6: 43255790-43255790 on Build GRCh38
3351477 c.853G>T p.D285Y Chr.6: 43255848-43255848 on Build GRCh38
6235012 c.798A>G p.S266S Chr.6: 43255793-43255793 on Build GRCh38
5186217 c.853G>A p.D285N Chr.6: 43255848-43255848 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751541280 Chr.6: 43255830-43255830 on Build GRCh38
rs1045031641 Chr.6: 43255960-43255960 on Build GRCh38
rs748781100 Chr.6: 43255866-43255866 on Build GRCh38
rs376062414 Chr.6: 43255790-43255790 on Build GRCh38
rs753849788 Chr.6: 43255860-43255860 on Build GRCh38
rs771579220 Chr.6: 43255888-43255888 on Build GRCh38
rs760804934 Chr.6: 43255742-43255742 on Build GRCh38
rs750196130 Chr.6: 43255844-43255844 on Build GRCh38
rs115138863 Chr.6: 43255933-43255933 on Build GRCh38
rs948028611 Chr.6: 43255948-43255948 on Build GRCh38
rs768099596 Chr.6: 43255878-43255878 on Build GRCh38
rs778392533 Chr.6: 43255880-43255880 on Build GRCh38
rs143865481 Chr.6: 43255829-43255829 on Build GRCh38
rs987154363 Chr.6: 43255957-43255957 on Build GRCh38
rs374636343 Chr.6: 43255927-43255927 on Build GRCh38
rs192229532 Chr.6: 43255865-43255865 on Build GRCh38
rs868215937 Chr.6: 43255752-43255752 on Build GRCh38
rs752786112 Chr.6: 43255819-43255819 on Build GRCh38
rs764229122 Chr.6: 43255905-43255905 on Build GRCh38
rs866322819 Chr.6: 43255971-43255971 on Build GRCh38
rs759851686 Chr.6: 43255893-43255893 on Build GRCh38
rs373096251 Chr.6: 43255767-43255767 on Build GRCh38
rs759748655 Chr.6: 43255768-43255768 on Build GRCh38
rs747576948 Chr.6: 43255881-43255881 on Build GRCh38
rs770012918 Chr.6: 43255895-43255895 on Build GRCh38
rs1024989378 Chr.6: 43255812-43255812 on Build GRCh38
rs775626073 Chr.6: 43255896-43255896 on Build GRCh38
rs368382639 Chr.6: 43255889-43255889 on Build GRCh38
rs781018619 Chr.6: 43255832-43255832 on Build GRCh38
rs979330391 Chr.6: 43255916-43255916 on Build GRCh38
rs917943022 Chr.6: 43255940-43255940 on Build GRCh38
rs370359466 Chr.6: 43255756-43255756 on Build GRCh38
rs763061957 Chr.6: 43255897-43255897 on Build GRCh38
rs967943020 Chr.6: 43255912-43255912 on Build GRCh38
rs371564967 Chr.6: 43255850-43255850 on Build GRCh38
rs536012213 Chr.6: 43255751-43255751 on Build GRCh38
rs887960206 Chr.6: 43255772-43255772 on Build GRCh38
rs923764407 Chr.6: 43255926-43255926 on Build GRCh38
rs761098675 Chr.6: 43255753-43255753 on Build GRCh38
rs911605058 Chr.6: 43255963-43255963 on Build GRCh38
rs34538060 Chr.6: 43255934-43255934 on Build GRCh38

View Full Product Details