Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 73704237-73704502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STX1A
Entrez Gene ID
6804
Gene Name
syntaxin 1A
Gene Aliases
HPC-1, P35-1, STX1, SYN1A
UniGene
Hs.647024
Gene Chromosome Location
Chr.7: 73699205-73719702 on Build GRCh38
Transcripts
NM_001165903.1 NM_004603.3 XM_017012567.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5463655 c.284-5C>A p.? Chr.7: 73704428-73704428 on Build GRCh38
1496662 c.363C>T p.S121S Chr.7: 73704251-73704251 on Build GRCh38
1266992 c.304C>T p.Q102* Chr.7: 73704403-73704403 on Build GRCh38
5357991 c.369G>T p.L123L Chr.7: 73704245-73704245 on Build GRCh38
3882267 c.358-1G>A p.? Chr.7: 73704257-73704257 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200947866 Chr.7: 73704442-73704442 on Build GRCh38
rs782136591 Chr.7: 73704447-73704447 on Build GRCh38
rs782380823 Chr.7: 73704456-73704456 on Build GRCh38
rs782226371 Chr.7: 73704385-73704385 on Build GRCh38
rs781875405 Chr.7: 73704356-73704356 on Build GRCh38
rs781921432 Chr.7: 73704330-73704330 on Build GRCh38
rs782462786 Chr.7: 73704404-73704404 on Build GRCh38
rs782383660 Chr.7: 73704298-73704298 on Build GRCh38
rs781790212 Chr.7: 73704342-73704342 on Build GRCh38
rs1047498015 Chr.7: 73704391-73704391 on Build GRCh38
rs782656169 Chr.7: 73704435-73704435 on Build GRCh38
rs782666829 Chr.7: 73704267-73704267 on Build GRCh38
rs376015107 Chr.7: 73704283-73704283 on Build GRCh38
rs782556908 Chr.7: 73704281-73704281 on Build GRCh38
rs1002422872 Chr.7: 73704480-73704480 on Build GRCh38
rs930291949 Chr.7: 73704346-73704346 on Build GRCh38
rs782724276 Chr.7: 73704325-73704325 on Build GRCh38
rs782304133 Chr.7: 73704295-73704295 on Build GRCh38
rs782105828 Chr.7: 73704339-73704339 on Build GRCh38
rs781986200 Chr.7: 73704307-73704307 on Build GRCh38
rs782722066 Chr.7: 73704475-73704475 on Build GRCh38
rs1055769456 Chr.7: 73704482-73704482 on Build GRCh38
rs782244597 Chr.7: 73704273-73704273 on Build GRCh38
rs907617478 Chr.7: 73704393-73704393 on Build GRCh38
rs781959209 Chr.7: 73704296-73704296 on Build GRCh38
rs202235482 Chr.7: 73704443-73704443 on Build GRCh38
rs199566653 Chr.7: 73704259-73704259 on Build GRCh38
rs782731994 Chr.7: 73704345-73704345 on Build GRCh38
rs782433844 Chr.7: 73704431-73704431 on Build GRCh38
rs74921525 Chr.7: 73704471-73704471 on Build GRCh38
rs782041280 Chr.7: 73704469-73704469 on Build GRCh38
rs782715874 Chr.7: 73704318-73704318 on Build GRCh38
rs781895205 Chr.7: 73704368-73704368 on Build GRCh38
rs782560813 Chr.7: 73704383-73704383 on Build GRCh38
rs782236446 Chr.7: 73704409-73704409 on Build GRCh38
rs782158668 Chr.7: 73704315-73704315 on Build GRCh38
rs782810999 Chr.7: 73704365-73704365 on Build GRCh38
rs782657148 Chr.7: 73704406-73704406 on Build GRCh38
rs782517349 Chr.7: 73704360-73704360 on Build GRCh38
rs782446724 Chr.7: 73704377-73704377 on Build GRCh38
rs782470638 Chr.7: 73704262-73704262 on Build GRCh38

View Full Product Details