Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 1208763-1209026 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC6A19
Entrez Gene ID
340024
Gene Name
solute carrier family 6 member 19
Gene Aliases
B0AT1, HND
UniGene
Hs.481478
Gene Chromosome Location
Chr.5: 1201595-1225117 on Build GRCh38
Transcripts
NM_001003841.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
735928 c.326C>T p.P109L Chr.5: 1208869-1208869 on Build GRCh38
3776123 c.276C>T p.I92I Chr.5: 1208819-1208819 on Build GRCh38
2149317 c.284G>A p.R95Q Chr.5: 1208827-1208827 on Build GRCh38
3946786 c.277G>T p.G93W Chr.5: 1208820-1208820 on Build GRCh38
3607851 c.338G>A p.G113D Chr.5: 1208881-1208881 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1042488670 Chr.5: 1208902-1208902 on Build GRCh38
rs757679627 Chr.5: 1208820-1208820 on Build GRCh38
rs752229715 Chr.5: 1208914-1208914 on Build GRCh38
rs982400558 Chr.5: 1208852-1208852 on Build GRCh38
rs548147900 Chr.5: 1208916-1208916 on Build GRCh38
rs758937137 Chr.5: 1208886-1208886 on Build GRCh38
rs1027984236 Chr.5: 1208831-1208831 on Build GRCh38
rs200447881 Chr.5: 1208927-1208927 on Build GRCh38
rs373263559 Chr.5: 1208913-1208913 on Build GRCh38
rs370115150 Chr.5: 1208835-1208836 on Build GRCh38
rs774243050 Chr.5: 1208904-1208942 on Build GRCh38
rs768901084 Chr.5: 1208899-1208899 on Build GRCh38
rs1040604013 Chr.5: 1209000-1209000 on Build GRCh38
rs6865548 Chr.5: 1208901-1208901 on Build GRCh38
rs902426205 Chr.5: 1208805-1208805 on Build GRCh38
rs371759358 Chr.5: 1208813-1208813 on Build GRCh38
rs752147563 Chr.5: 1208934-1208934 on Build GRCh38
rs780877780 Chr.5: 1208815-1208815 on Build GRCh38
rs959925419 Chr.5: 1208972-1208972 on Build GRCh38
rs762963674 Chr.5: 1208845-1208845 on Build GRCh38
rs765742717 Chr.5: 1208846-1208846 on Build GRCh38
rs72709422 Chr.5: 1208948-1208948 on Build GRCh38
rs748425336 Chr.5: 1208907-1208907 on Build GRCh38
rs750826028 Chr.5: 1208872-1208872 on Build GRCh38
rs201936518 Chr.5: 1208827-1208827 on Build GRCh38
rs1012357083 Chr.5: 1208973-1208973 on Build GRCh38
rs60992377 Chr.5: 1208849-1208849 on Build GRCh38
rs191798547 Chr.5: 1209004-1209004 on Build GRCh38
rs114684753 Chr.5: 1208870-1208870 on Build GRCh38
rs769418030 Chr.5: 1208908-1208908 on Build GRCh38
rs944779668 Chr.5: 1208989-1208989 on Build GRCh38
rs777046481 Chr.5: 1208835-1208835 on Build GRCh38
rs557347561 Chr.5: 1208962-1208962 on Build GRCh38
rs575201026 Chr.5: 1208965-1208965 on Build GRCh38
rs915242991 Chr.5: 1208900-1208900 on Build GRCh38
rs112140006 Chr.5: 1208952-1208952 on Build GRCh38
rs201010661 Chr.5: 1208828-1208828 on Build GRCh38
rs369605197 Chr.5: 1208836-1208836 on Build GRCh38
rs537565167 Chr.5: 1208912-1208912 on Build GRCh38
rs557785841 Chr.5: 1208979-1208979 on Build GRCh38
rs750788916 Chr.5: 1208925-1208925 on Build GRCh38
rs937924348 Chr.5: 1208895-1208895 on Build GRCh38
rs146678323 Chr.5: 1208819-1208819 on Build GRCh38
rs564753776 Chr.5: 1208858-1208858 on Build GRCh38
rs146085736 Chr.5: 1208897-1208897 on Build GRCh38
rs761761040 Chr.5: 1208798-1208798 on Build GRCh38
rs771032164 Chr.5: 1208867-1208867 on Build GRCh38
rs552258859 Chr.5: 1208935-1208935 on Build GRCh38
rs944586830 Chr.5: 1208923-1208923 on Build GRCh38
rs759094266 Chr.5: 1208854-1208854 on Build GRCh38
rs747155573 Chr.5: 1208832-1208832 on Build GRCh38
rs767124586 Chr.5: 1208868-1208868 on Build GRCh38
rs752378013 Chr.5: 1208869-1208869 on Build GRCh38
rs1040293999 Chr.5: 1208943-1208943 on Build GRCh38
rs967742224 Chr.5: 1208986-1208986 on Build GRCh38
rs772975003 Chr.5: 1208844-1208844 on Build GRCh38
rs369819680 Chr.5: 1208978-1208978 on Build GRCh38
rs897163245 Chr.5: 1208959-1208959 on Build GRCh38
rs765066805 Chr.5: 1208801-1208801 on Build GRCh38
rs111708279 Chr.5: 1208956-1208956 on Build GRCh38
rs765936901 Chr.5: 1208807-1208807 on Build GRCh38
rs368177876 Chr.5: 1208806-1208806 on Build GRCh38
rs376995967 Chr.5: 1208814-1208814 on Build GRCh38
rs369767035 Chr.5: 1208909-1208909 on Build GRCh38
rs768989277 Chr.5: 1208833-1208833 on Build GRCh38
rs747065876 Chr.5: 1208898-1208898 on Build GRCh38
rs139182948 Chr.5: 1208826-1208826 on Build GRCh38
rs1005660014 Chr.5: 1208954-1208954 on Build GRCh38
rs769662523 Chr.5: 1208841-1208841 on Build GRCh38
rs867695283 Chr.5: 1208825-1208825 on Build GRCh38
rs750418535 Chr.5: 1208802-1208802 on Build GRCh38
rs571058594 Chr.5: 1208942-1208942 on Build GRCh38
rs921882769 Chr.5: 1208947-1208947 on Build GRCh38
rs547020951 Chr.5: 1208951-1208951 on Build GRCh38
rs1035565614 Chr.5: 1208864-1208864 on Build GRCh38
rs760376373 Chr.5: 1208915-1208915 on Build GRCh38
rs563106678 Chr.5: 1208926-1208926 on Build GRCh38

View Full Product Details