Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 140563127-140563400 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UCP1
Entrez Gene ID
7350
Gene Name
uncoupling protein 1
Gene Aliases
SLC25A7, UCP
UniGene
Hs.249211
Gene Chromosome Location
Chr.4: 140555770-140568914 on Build GRCh38
Transcripts
NM_021833.4 XM_005263206.3 XM_011532228.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4158815 c.465G>A p.T155T Chr.4: 140563379-140563379 on Build GRCh38
4963202 c.586G>T p.D196Y Chr.4: 140563152-140563152 on Build GRCh38
1427249 c.517C>A p.L173I Chr.4: 140563327-140563327 on Build GRCh38
3125772 c.527-7T>C p.? Chr.4: 140563218-140563218 on Build GRCh38
6816162 c.512C>T p.T171M Chr.4: 140563332-140563332 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747261064 Chr.4: 140563355-140563355 on Build GRCh38
rs1010392553 Chr.4: 140563187-140563187 on Build GRCh38
rs200389729 Chr.4: 140563365-140563365 on Build GRCh38
rs867228230 Chr.4: 140563322-140563322 on Build GRCh38
rs374147979 Chr.4: 140563278-140563278 on Build GRCh38
rs778088952 Chr.4: 140563235-140563235 on Build GRCh38
rs1023720944 Chr.4: 140563213-140563214 on Build GRCh38
rs577520173 Chr.4: 140563209-140563209 on Build GRCh38
rs769644397 Chr.4: 140563270-140563270 on Build GRCh38
rs200874530 Chr.4: 140563215-140563215 on Build GRCh38
rs548319096 Chr.4: 140563338-140563338 on Build GRCh38
rs769473091 Chr.4: 140563364-140563364 on Build GRCh38
rs111460010 Chr.4: 140563218-140563218 on Build GRCh38
rs756530707 Chr.4: 140563229-140563229 on Build GRCh38
rs188223067 Chr.4: 140563197-140563197 on Build GRCh38
rs772226264 Chr.4: 140563250-140563250 on Build GRCh38
rs763504112 Chr.4: 140563352-140563354 on Build GRCh38
rs749324285 Chr.4: 140563156-140563156 on Build GRCh38
rs771967103 Chr.4: 140563216-140563217 on Build GRCh38
rs765046420 Chr.4: 140563208-140563208 on Build GRCh38
rs909901649 Chr.4: 140563264-140563264 on Build GRCh38
rs767034123 Chr.4: 140563294-140563294 on Build GRCh38
rs781596160 Chr.4: 140563226-140563226 on Build GRCh38
rs74932109 Chr.4: 140563214-140563214 on Build GRCh38
rs765974159 Chr.4: 140563285-140563285 on Build GRCh38
rs527369932 Chr.4: 140563176-140563178 on Build GRCh38
rs140830198 Chr.4: 140563343-140563343 on Build GRCh38
rs150152405 Chr.4: 140563371-140563371 on Build GRCh38
rs761468229 Chr.4: 140563268-140563268 on Build GRCh38
rs552377264 Chr.4: 140563213-140563213 on Build GRCh38
rs559928395 Chr.4: 140563218-140563218 on Build GRCh38
rs192333501 Chr.4: 140563379-140563379 on Build GRCh38
rs1032087150 Chr.4: 140563165-140563165 on Build GRCh38
rs770251704 Chr.4: 140563154-140563154 on Build GRCh38
rs753041529 Chr.4: 140563227-140563227 on Build GRCh38
rs746686043 Chr.4: 140563217-140563218 on Build GRCh38
rs374031496 Chr.4: 140563223-140563223 on Build GRCh38
rs544110432 Chr.4: 140563155-140563155 on Build GRCh38
rs202215906 Chr.4: 140563186-140563186 on Build GRCh38
rs531672425 Chr.4: 140563380-140563380 on Build GRCh38
rs778934896 Chr.4: 140563339-140563339 on Build GRCh38
rs367909350 Chr.4: 140563246-140563246 on Build GRCh38
rs751020946 Chr.4: 140563238-140563238 on Build GRCh38
rs757707004 Chr.4: 140563244-140563244 on Build GRCh38
rs1000573339 Chr.4: 140563164-140563164 on Build GRCh38
rs774495998 Chr.4: 140563160-140563160 on Build GRCh38
rs942721402 Chr.4: 140563265-140563265 on Build GRCh38
rs759194010 Chr.4: 140563287-140563287 on Build GRCh38
rs200354766 Chr.4: 140563190-140563190 on Build GRCh38
rs756371993 Chr.4: 140563328-140563328 on Build GRCh38
rs770997395 Chr.4: 140563242-140563242 on Build GRCh38
rs371409039 Chr.4: 140563262-140563262 on Build GRCh38
rs763570956 Chr.4: 140563203-140563203 on Build GRCh38
rs776236480 Chr.4: 140563218-140563219 on Build GRCh38
rs759825652 Chr.4: 140563182-140563182 on Build GRCh38
rs763050237 Chr.4: 140563284-140563284 on Build GRCh38
rs757923988 Chr.4: 140563217-140563217 on Build GRCh38
rs745628003 Chr.4: 140563214-140563214 on Build GRCh38
rs751162311 Chr.4: 140563286-140563286 on Build GRCh38
rs376167170 Chr.4: 140563158-140563158 on Build GRCh38
rs866125734 Chr.4: 140563359-140563359 on Build GRCh38
rs778111188 Chr.4: 140563330-140563330 on Build GRCh38
rs147603400 Chr.4: 140563342-140563342 on Build GRCh38
rs748672923 Chr.4: 140563152-140563152 on Build GRCh38
rs752414301 Chr.4: 140563302-140563302 on Build GRCh38
rs754333261 Chr.4: 140563331-140563331 on Build GRCh38

View Full Product Details