Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 41256886-41257119 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UCHL1
Entrez Gene ID
7345
Gene Name
ubiquitin C-terminal hydrolase L1
Gene Aliases
HEL-117, HEL-S-53, NDGOA, PARK5, PGP 9.5, PGP9.5, PGP95, Uch-L1
UniGene
Hs.518731
Gene Chromosome Location
Chr.4: 41256881-41268429 on Build GRCh38
Transcripts
NM_004181.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3775844 c.14C>G p.P5R Chr.4: 41256990-41256990 on Build GRCh38
1753727 c.15G>A p.P5P Chr.4: 41256991-41256991 on Build GRCh38
1618817 c.33+6C>T p.? Chr.4: 41257015-41257015 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs762425804 Chr.4: 41257006-41257006 on Build GRCh38
rs755271457 Chr.4: 41257090-41257091 on Build GRCh38
rs757111252 Chr.4: 41256970-41256970 on Build GRCh38
rs756428586 Chr.4: 41256964-41256964 on Build GRCh38
rs750684470 Chr.4: 41256939-41256939 on Build GRCh38
rs766696958 Chr.4: 41256965-41256965 on Build GRCh38
rs769034726 Chr.4: 41256998-41256998 on Build GRCh38
rs755648455 Chr.4: 41257067-41257067 on Build GRCh38
rs781003024 Chr.4: 41257060-41257060 on Build GRCh38
rs375794773 Chr.4: 41256927-41256927 on Build GRCh38
rs577696101 Chr.4: 41256918-41256918 on Build GRCh38
rs778811044 Chr.4: 41256945-41256945 on Build GRCh38
rs9321 Chr.4: 41256961-41256961 on Build GRCh38
rs745572268 Chr.4: 41256954-41256954 on Build GRCh38
rs769288409 Chr.4: 41256982-41256982 on Build GRCh38
rs746517688 Chr.4: 41256923-41256923 on Build GRCh38
rs766500601 Chr.4: 41257035-41257035 on Build GRCh38
rs1041476078 Chr.4: 41257000-41257000 on Build GRCh38
rs752001613 Chr.4: 41256930-41256930 on Build GRCh38
rs372590899 Chr.4: 41257056-41257056 on Build GRCh38
rs756271669 Chr.4: 41256944-41256944 on Build GRCh38
rs774679952 Chr.4: 41257001-41257001 on Build GRCh38
rs747436842 Chr.4: 41257056-41257056 on Build GRCh38
rs113713555 Chr.4: 41257049-41257049 on Build GRCh38
rs748992332 Chr.4: 41257081-41257081 on Build GRCh38
rs886059407 Chr.4: 41257014-41257014 on Build GRCh38
rs747954734 Chr.4: 41256946-41256946 on Build GRCh38
rs757276311 Chr.4: 41257057-41257057 on Build GRCh38
rs777866474 Chr.4: 41256950-41256950 on Build GRCh38
rs746877403 Chr.4: 41256951-41256951 on Build GRCh38
rs775798907 Chr.4: 41256928-41256928 on Build GRCh38
rs562809259 Chr.4: 41257028-41257028 on Build GRCh38
rs11556271 Chr.4: 41256953-41256953 on Build GRCh38
rs767689458 Chr.4: 41256962-41256962 on Build GRCh38
rs371943957 Chr.4: 41256926-41256926 on Build GRCh38
rs754144624 Chr.4: 41257043-41257043 on Build GRCh38
rs769324047 Chr.4: 41256957-41256957 on Build GRCh38
rs759809182 Chr.4: 41257048-41257048 on Build GRCh38
rs1054299077 Chr.4: 41257050-41257050 on Build GRCh38
rs750305902 Chr.4: 41257062-41257062 on Build GRCh38
rs901082145 Chr.4: 41256931-41256931 on Build GRCh38
rs1040707634 Chr.4: 41256985-41256985 on Build GRCh38
rs924428961 Chr.4: 41256925-41256925 on Build GRCh38
rs751596305 Chr.4: 41256969-41256969 on Build GRCh38
rs779486680 Chr.4: 41257071-41257071 on Build GRCh38
rs397515634 Chr.4: 41256996-41256996 on Build GRCh38
rs745355343 Chr.4: 41256978-41256978 on Build GRCh38
rs201952662 Chr.4: 41256949-41256949 on Build GRCh38
rs991106482 Chr.4: 41256990-41256990 on Build GRCh38
rs775888515 Chr.4: 41256959-41256959 on Build GRCh38
rs958992248 Chr.4: 41256972-41256972 on Build GRCh38
rs780819970 Chr.4: 41256974-41256974 on Build GRCh38
rs375157748 Chr.4: 41257013-41257013 on Build GRCh38
rs200302762 Chr.4: 41256966-41256966 on Build GRCh38
rs201864413 Chr.4: 41256992-41256992 on Build GRCh38
rs779681524 Chr.4: 41256983-41256983 on Build GRCh38
rs949837746 Chr.4: 41256905-41256905 on Build GRCh38
rs6832687 Chr.4: 41257096-41257096 on Build GRCh38
rs903038210 Chr.4: 41256916-41256916 on Build GRCh38
rs201100258 Chr.4: 41256906-41256906 on Build GRCh38
rs757358332 Chr.4: 41256933-41256933 on Build GRCh38
rs367935445 Chr.4: 41256960-41256960 on Build GRCh38
rs767459303 Chr.4: 41256937-41256937 on Build GRCh38
rs763516024 Chr.4: 41256929-41256929 on Build GRCh38

View Full Product Details