Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 49864642-49864895 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC9A8
Entrez Gene ID
23315
Gene Name
solute carrier family 9 member A8
Gene Aliases
NHE-8, NHE8
UniGene
Hs.444202
Gene Chromosome Location
Chr.20: 49812713-49892242 on Build GRCh38
Transcripts
NM_001260491.1 NM_015266.2 NR_048537.1 NR_048538.1 NR_048539.1 NR_048540.1 XM_006723756.1 XM_011528736.1 XM_011528737.1 XM_011528738.1 XM_011528739.1 XM_011528740.1 XM_011528741.1 XM_011528742.1 XM_011528743.2 XM_011528744.2 XM_017027754.1 XM_017027755.1 XM_017027756.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6484864 c.977A>G p.Y326C Chr.20: 49864815-49864815 on Build GRCh38
3748754 c.942G>A p.E314E Chr.20: 49864780-49864780 on Build GRCh38
3748755 c.894G>A p.E298E Chr.20: 49864780-49864780 on Build GRCh38
350215 c.950C>A p.S317* Chr.20: 49864836-49864836 on Build GRCh38
7194563 c.935C>A p.S312Y Chr.20: 49864773-49864773 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762856999 Chr.20: 49864733-49864733 on Build GRCh38
rs759363309 Chr.20: 49864739-49864739 on Build GRCh38
rs758807708 Chr.20: 49864805-49864805 on Build GRCh38
rs949708399 Chr.20: 49864810-49864810 on Build GRCh38
rs768724377 Chr.20: 49864853-49864853 on Build GRCh38
rs753429040 Chr.20: 49864774-49864774 on Build GRCh38
rs776927808 Chr.20: 49864709-49864709 on Build GRCh38
rs747521521 Chr.20: 49864763-49864763 on Build GRCh38
rs201073525 Chr.20: 49864741-49864741 on Build GRCh38
rs146050863 Chr.20: 49864708-49864708 on Build GRCh38
rs936227315 Chr.20: 49864720-49864720 on Build GRCh38
rs765677672 Chr.20: 49864707-49864717 on Build GRCh38
rs1042012230 Chr.20: 49864702-49864702 on Build GRCh38
rs369856632 Chr.20: 49864787-49864787 on Build GRCh38
rs746412731 Chr.20: 49864802-49864802 on Build GRCh38
rs757673895 Chr.20: 49864783-49864783 on Build GRCh38
rs558335310 Chr.20: 49864872-49864872 on Build GRCh38
rs760042489 Chr.20: 49864702-49864703 on Build GRCh38
rs138829825 Chr.20: 49864767-49864767 on Build GRCh38
rs759330189 Chr.20: 49864875-49864875 on Build GRCh38
rs751616721 Chr.20: 49864738-49864738 on Build GRCh38
rs748378122 Chr.20: 49864859-49864859 on Build GRCh38
rs774268818 Chr.20: 49864874-49864874 on Build GRCh38
rs766264617 Chr.20: 49864734-49864734 on Build GRCh38
rs780562016 Chr.20: 49864808-49864808 on Build GRCh38
rs188598081 Chr.20: 49864847-49864847 on Build GRCh38
rs372415486 Chr.20: 49864707-49864707 on Build GRCh38
rs772259617 Chr.20: 49864697-49864697 on Build GRCh38
rs775399242 Chr.20: 49864704-49864704 on Build GRCh38
rs746357603 Chr.20: 49864690-49864690 on Build GRCh38
rs897680353 Chr.20: 49864830-49864830 on Build GRCh38
rs752174664 Chr.20: 49864742-49864742 on Build GRCh38
rs763653482 Chr.20: 49864768-49864768 on Build GRCh38
rs776856026 Chr.20: 49864697-49864698 on Build GRCh38

View Full Product Details