Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40745027-40745300 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf54
Entrez Gene ID
284325
Gene Name
chromosome 19 open reading frame 54
Gene Aliases
-
UniGene
Hs.585105
Gene Chromosome Location
Chr.19: 40740856-40750713 on Build GRCh38
Transcripts
NM_198476.3 XM_005258776.4 XM_005258778.4 XM_006723152.2 XM_006723153.2 XM_011526774.2 XM_011526775.1 XM_011526776.2 XM_011526777.2 XM_011526778.2 XM_011526779.2 XM_011526782.2 XM_011526783.2 XM_011526784.2 XM_011526785.2 XM_011526786.2 XM_017026620.1 XM_017026621.1 XM_017026622.1 XM_017026623.1 XM_017026624.1 XM_017026625.1 XM_017026626.1 XM_017026627.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5741594 c.341G>A p.R114Q Chr.19: 40745181-40745181 on Build GRCh38
4489898 c.354C>T p.F118F Chr.19: 40745168-40745168 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs375442763 Chr.19: 40745115-40745115 on Build GRCh38
rs900380100 Chr.19: 40745177-40745177 on Build GRCh38
rs776146552 Chr.19: 40745086-40745086 on Build GRCh38
rs957990465 Chr.19: 40745221-40745221 on Build GRCh38
rs762818408 Chr.19: 40745214-40745214 on Build GRCh38
rs868769385 Chr.19: 40745130-40745130 on Build GRCh38
rs749005034 Chr.19: 40745076-40745076 on Build GRCh38
rs887023616 Chr.19: 40745170-40745170 on Build GRCh38
rs889305856 Chr.19: 40745053-40745053 on Build GRCh38
rs186635253 Chr.19: 40745235-40745235 on Build GRCh38
rs372150560 Chr.19: 40745211-40745211 on Build GRCh38
rs12151086 Chr.19: 40745254-40745254 on Build GRCh38
rs886218956 Chr.19: 40745136-40745136 on Build GRCh38
rs571137325 Chr.19: 40745161-40745161 on Build GRCh38
rs199729288 Chr.19: 40745193-40745193 on Build GRCh38
rs898847124 Chr.19: 40745119-40745119 on Build GRCh38
rs547873352 Chr.19: 40745212-40745212 on Build GRCh38
rs2369302 Chr.19: 40745275-40745275 on Build GRCh38
rs776875611 Chr.19: 40745181-40745181 on Build GRCh38
rs745746569 Chr.19: 40745127-40745127 on Build GRCh38
rs368675093 Chr.19: 40745106-40745106 on Build GRCh38
rs772209283 Chr.19: 40745158-40745158 on Build GRCh38
rs745372979 Chr.19: 40745118-40745118 on Build GRCh38
rs768729264 Chr.19: 40745180-40745180 on Build GRCh38
rs930367010 Chr.19: 40745129-40745129 on Build GRCh38
rs138009799 Chr.19: 40745277-40745277 on Build GRCh38
rs762110015 Chr.19: 40745182-40745182 on Build GRCh38
rs980827592 Chr.19: 40745194-40745194 on Build GRCh38
rs370320401 Chr.19: 40745114-40745114 on Build GRCh38
rs995139230 Chr.19: 40745153-40745153 on Build GRCh38
rs562463349 Chr.19: 40745186-40745186 on Build GRCh38

View Full Product Details