Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 16792813-16793056 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGR2
Entrez Gene ID
10551
Gene Name
anterior gradient 2, protein disulphide isomerase family member
Gene Aliases
AG2, GOB-4, HAG-2, HEL-S-116, PDIA17, XAG-2
UniGene
Hs.530009
Gene Chromosome Location
Chr.7: 16791811-16805114 on Build GRCh38
Transcripts
NM_006408.3 XM_005249581.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5467092 c.497A>C p.K166T Chr.7: 16792939-16792939 on Build GRCh38
236964 c.479-5A>G p.? Chr.7: 16792962-16792962 on Build GRCh38
3636919 c.492G>A p.M164I Chr.7: 16792944-16792944 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs530285752 Chr.7: 16792985-16792985 on Build GRCh38
rs902896099 Chr.7: 16792930-16792930 on Build GRCh38
rs762996070 Chr.7: 16792868-16792868 on Build GRCh38
rs541219367 Chr.7: 16792906-16792906 on Build GRCh38
rs1051869 Chr.7: 16792851-16792851 on Build GRCh38
rs764986074 Chr.7: 16792997-16792997 on Build GRCh38
rs777605755 Chr.7: 16792961-16792961 on Build GRCh38
rs143032356 Chr.7: 16792910-16792910 on Build GRCh38
rs747674876 Chr.7: 16792956-16792956 on Build GRCh38
rs371917010 Chr.7: 16792984-16792984 on Build GRCh38
rs573471377 Chr.7: 16792857-16792857 on Build GRCh38
rs775748089 Chr.7: 16792967-16792967 on Build GRCh38
rs745671356 Chr.7: 16793009-16793009 on Build GRCh38
rs775599522 Chr.7: 16792866-16792866 on Build GRCh38
rs768027549 Chr.7: 16792991-16792991 on Build GRCh38
rs374504036 Chr.7: 16792963-16792963 on Build GRCh38
rs754532797 Chr.7: 16792920-16792920 on Build GRCh38
rs76194340 Chr.7: 16792999-16792999 on Build GRCh38
rs560550442 Chr.7: 16792882-16792882 on Build GRCh38
rs892787716 Chr.7: 16792844-16792844 on Build GRCh38
rs767921752 Chr.7: 16792876-16792879 on Build GRCh38
rs540529208 Chr.7: 16792846-16792846 on Build GRCh38
rs753474844 Chr.7: 16792911-16792911 on Build GRCh38
rs772072049 Chr.7: 16792960-16792960 on Build GRCh38
rs756465833 Chr.7: 16792900-16792900 on Build GRCh38
rs750731771 Chr.7: 16792898-16792898 on Build GRCh38
rs914912032 Chr.7: 16792863-16792863 on Build GRCh38
rs900117439 Chr.7: 16792998-16792998 on Build GRCh38
rs1802157 Chr.7: 16792873-16792873 on Build GRCh38
rs763224709 Chr.7: 16792980-16792980 on Build GRCh38
rs142501977 Chr.7: 16792995-16792995 on Build GRCh38
rs374150474 Chr.7: 16792992-16792992 on Build GRCh38
rs770021368 Chr.7: 16792862-16792862 on Build GRCh38
rs368448343 Chr.7: 16792966-16792966 on Build GRCh38
rs778625347 Chr.7: 16792936-16792936 on Build GRCh38
rs760395687 Chr.7: 16792858-16792858 on Build GRCh38
rs372693759 Chr.7: 16792996-16792996 on Build GRCh38
rs780903230 Chr.7: 16793007-16793007 on Build GRCh38
rs369355847 Chr.7: 16792854-16792854 on Build GRCh38
rs148204661 Chr.7: 16792950-16792950 on Build GRCh38
rs762361024 Chr.7: 16792881-16792881 on Build GRCh38
rs767923511 Chr.7: 16792885-16792885 on Build GRCh38
rs999650664 Chr.7: 16792945-16792945 on Build GRCh38
rs192581998 Chr.7: 16793000-16793000 on Build GRCh38
rs372290890 Chr.7: 16793011-16793011 on Build GRCh38
rs746956940 Chr.7: 16793005-16793005 on Build GRCh38
rs750452130 Chr.7: 16792898-16792898 on Build GRCh38
rs1041457820 Chr.7: 16792848-16792850 on Build GRCh38
rs752053196 Chr.7: 16792878-16792878 on Build GRCh38
rs923088522 Chr.7: 16793020-16793020 on Build GRCh38

View Full Product Details