Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 168239018-168239279 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SFT2D2
Entrez Gene ID
375035
Gene Name
SFT2 domain containing 2
Gene Aliases
UNQ512, dJ747L4.C1.2
UniGene
Hs.645435
Gene Chromosome Location
Chr.1: 168226017-168242850 on Build GRCh38
Transcripts
NM_199344.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
677300 c.420C>T p.S140S Chr.1: 168239137-168239137 on Build GRCh38
4024870 c.433C>T p.P145S Chr.1: 168239150-168239150 on Build GRCh38
6459580 c.421C>T p.L141F Chr.1: 168239138-168239138 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs370350839 Chr.1: 168239092-168239092 on Build GRCh38
rs948976356 Chr.1: 168239109-168239113 on Build GRCh38
rs1055183186 Chr.1: 168239143-168239143 on Build GRCh38
rs756873866 Chr.1: 168239181-168239181 on Build GRCh38
rs908672604 Chr.1: 168239076-168239076 on Build GRCh38
rs185866018 Chr.1: 168239196-168239196 on Build GRCh38
rs774802158 Chr.1: 168239058-168239058 on Build GRCh38
rs946659137 Chr.1: 168239208-168239208 on Build GRCh38
rs781711278 Chr.1: 168239108-168239108 on Build GRCh38
rs1017245584 Chr.1: 168239244-168239244 on Build GRCh38
rs536792918 Chr.1: 168239090-168239090 on Build GRCh38
rs752395361 Chr.1: 168239087-168239087 on Build GRCh38
rs761746572 Chr.1: 168239148-168239148 on Build GRCh38
rs1046418542 Chr.1: 168239127-168239127 on Build GRCh38
rs749428498 Chr.1: 168239139-168239139 on Build GRCh38
rs773060458 Chr.1: 168239168-168239169 on Build GRCh38
rs770978777 Chr.1: 168239142-168239142 on Build GRCh38
rs748408063 Chr.1: 168239130-168239130 on Build GRCh38
rs12142742 Chr.1: 168239070-168239070 on Build GRCh38
rs199565250 Chr.1: 168239164-168239164 on Build GRCh38
rs372749526 Chr.1: 168239079-168239079 on Build GRCh38
rs529443245 Chr.1: 168239136-168239136 on Build GRCh38
rs907338330 Chr.1: 168239131-168239131 on Build GRCh38
rs764986217 Chr.1: 168239247-168239247 on Build GRCh38
rs373943661 Chr.1: 168239185-168239185 on Build GRCh38
rs373049032 Chr.1: 168239099-168239099 on Build GRCh38
rs375226598 Chr.1: 168239063-168239063 on Build GRCh38
rs759502669 Chr.1: 168239152-168239152 on Build GRCh38
rs764859181 Chr.1: 168239195-168239195 on Build GRCh38
rs767375956 Chr.1: 168239153-168239153 on Build GRCh38
rs370765463 Chr.1: 168239175-168239175 on Build GRCh38
rs774367746 Chr.1: 168239149-168239149 on Build GRCh38
rs376045742 Chr.1: 168239163-168239163 on Build GRCh38
rs773304727 Chr.1: 168239138-168239138 on Build GRCh38
rs747493031 Chr.1: 168239107-168239107 on Build GRCh38
rs763871684 Chr.1: 168239174-168239174 on Build GRCh38
rs774773715 Chr.1: 168239245-168239245 on Build GRCh38

View Full Product Details