Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 82365614-82365849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TPBG
Entrez Gene ID
7162
Gene Name
trophoblast glycoprotein
Gene Aliases
5T4, 5T4AG, M6P1, WAIF1
UniGene
Hs.71947
Gene Chromosome Location
Chr.6: 82363206-82370828 on Build GRCh38
Transcripts
NM_001166392.1 NM_006670.4 XM_011536097.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5710241 c.813G>C p.K271N Chr.6: 82365774-82365774 on Build GRCh38
1329534 c.714C>T p.H238H Chr.6: 82365675-82365675 on Build GRCh38
238339 c.735G>A p.S245S Chr.6: 82365696-82365696 on Build GRCh38
5567795 c.725G>C p.S242T Chr.6: 82365686-82365686 on Build GRCh38
743015 c.702C>T p.P234P Chr.6: 82365663-82365663 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199757455 Chr.6: 82365760-82365760 on Build GRCh38
rs371527625 Chr.6: 82365766-82365766 on Build GRCh38
rs138578663 Chr.6: 82365725-82365725 on Build GRCh38
rs377740252 Chr.6: 82365685-82365685 on Build GRCh38
rs1029700020 Chr.6: 82365672-82365672 on Build GRCh38
rs760957275 Chr.6: 82365673-82365673 on Build GRCh38
rs780665666 Chr.6: 82365810-82365810 on Build GRCh38
rs776844600 Chr.6: 82365764-82365764 on Build GRCh38
rs374149146 Chr.6: 82365710-82365710 on Build GRCh38
rs777352855 Chr.6: 82365706-82365706 on Build GRCh38
rs771258090 Chr.6: 82365720-82365720 on Build GRCh38
rs931897460 Chr.6: 82365785-82365785 on Build GRCh38
rs750216770 Chr.6: 82365678-82365678 on Build GRCh38
rs752239077 Chr.6: 82365774-82365774 on Build GRCh38
rs749642884 Chr.6: 82365644-82365645 on Build GRCh38
rs760787559 Chr.6: 82365757-82365757 on Build GRCh38
rs759537182 Chr.6: 82365660-82365660 on Build GRCh38
rs200883951 Chr.6: 82365795-82365795 on Build GRCh38
rs772728489 Chr.6: 82365807-82365807 on Build GRCh38
rs772787081 Chr.6: 82365793-82365794 on Build GRCh38
rs974744156 Chr.6: 82365709-82365709 on Build GRCh38
rs978273505 Chr.6: 82365641-82365641 on Build GRCh38
rs145977230 Chr.6: 82365695-82365695 on Build GRCh38
rs376116704 Chr.6: 82365767-82365767 on Build GRCh38
rs767398273 Chr.6: 82365771-82365771 on Build GRCh38
rs1054762709 Chr.6: 82365820-82365820 on Build GRCh38
rs771730945 Chr.6: 82365774-82365774 on Build GRCh38
rs753517966 Chr.6: 82365781-82365781 on Build GRCh38
rs759433568 Chr.6: 82365769-82365769 on Build GRCh38
rs774860674 Chr.6: 82365654-82365654 on Build GRCh38
rs62431321 Chr.6: 82365751-82365751 on Build GRCh38
rs201434343 Chr.6: 82365788-82365788 on Build GRCh38
rs924451033 Chr.6: 82365772-82365772 on Build GRCh38
rs775912512 Chr.6: 82365663-82365663 on Build GRCh38
rs747088316 Chr.6: 82365813-82365813 on Build GRCh38
rs755558338 Chr.6: 82365776-82365776 on Build GRCh38
rs372123730 Chr.6: 82365747-82365747 on Build GRCh38
rs138970644 Chr.6: 82365642-82365642 on Build GRCh38
rs766180931 Chr.6: 82365768-82365768 on Build GRCh38
rs768796802 Chr.6: 82365818-82365818 on Build GRCh38
rs201599531 Chr.6: 82365696-82365696 on Build GRCh38
rs142111937 Chr.6: 82365691-82365691 on Build GRCh38
rs187936619 Chr.6: 82365649-82365649 on Build GRCh38
rs777307476 Chr.6: 82365777-82365777 on Build GRCh38
rs765163936 Chr.6: 82365676-82365676 on Build GRCh38
rs749625694 Chr.6: 82365711-82365711 on Build GRCh38
rs776773661 Chr.6: 82365819-82365819 on Build GRCh38
rs200075724 Chr.6: 82365697-82365697 on Build GRCh38
rs368363343 Chr.6: 82365717-82365717 on Build GRCh38
rs141575389 Chr.6: 82365730-82365730 on Build GRCh38
rs767699453 Chr.6: 82365662-82365662 on Build GRCh38
rs772327360 Chr.6: 82365742-82365742 on Build GRCh38
rs754280858 Chr.6: 82365793-82365793 on Build GRCh38
rs746411153 Chr.6: 82365803-82365803 on Build GRCh38

View Full Product Details