Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 169796237-169796462 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC34
Entrez Gene ID
151827
Gene Name
leucine rich repeat containing 34
Gene Aliases
-
UniGene
Hs.591289
Gene Chromosome Location
Chr.3: 169793006-169812904 on Build GRCh38
Transcripts
NM_001172779.1 NM_001172780.1 NM_153353.4 XM_006713508.3 XM_011512442.2 XM_017005746.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5834605 c.1020C>G p.N340K Chr.3: 169796258-169796258 on Build GRCh38
3203836 c.780A>C p.K260N Chr.3: 169796363-169796363 on Build GRCh38
7181353 c.791A>C p.D264A Chr.3: 169796352-169796352 on Build GRCh38
4629485 c.863G>C p.R288T Chr.3: 169796280-169796280 on Build GRCh38
6577860 c.842T>C p.V281A Chr.3: 169796301-169796301 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753932069 Chr.3: 169796388-169796388 on Build GRCh38
rs148256393 Chr.3: 169796291-169796291 on Build GRCh38
rs766041615 Chr.3: 169796330-169796330 on Build GRCh38
rs141628545 Chr.3: 169796312-169796312 on Build GRCh38
rs774644483 Chr.3: 169796355-169796355 on Build GRCh38
rs138701364 Chr.3: 169796380-169796380 on Build GRCh38
rs1003188387 Chr.3: 169796410-169796410 on Build GRCh38
rs762557836 Chr.3: 169796319-169796319 on Build GRCh38
rs146400850 Chr.3: 169796285-169796289 on Build GRCh38
rs758414790 Chr.3: 169796288-169796288 on Build GRCh38
rs746024128 Chr.3: 169796413-169796413 on Build GRCh38
rs888641296 Chr.3: 169796340-169796341 on Build GRCh38
rs377368072 Chr.3: 169796293-169796293 on Build GRCh38
rs756574854 Chr.3: 169796376-169796376 on Build GRCh38
rs201312558 Chr.3: 169796361-169796361 on Build GRCh38
rs756202632 Chr.3: 169796264-169796264 on Build GRCh38
rs769215619 Chr.3: 169796315-169796315 on Build GRCh38
rs777861751 Chr.3: 169796277-169796277 on Build GRCh38
rs370011461 Chr.3: 169796289-169796293 on Build GRCh38
rs753226504 Chr.3: 169796373-169796373 on Build GRCh38
rs776717065 Chr.3: 169796310-169796310 on Build GRCh38
rs757482546 Chr.3: 169796397-169796397 on Build GRCh38
rs932800860 Chr.3: 169796308-169796308 on Build GRCh38
rs911784486 Chr.3: 169796265-169796265 on Build GRCh38
rs921027967 Chr.3: 169796296-169796296 on Build GRCh38
rs202031258 Chr.3: 169796280-169796280 on Build GRCh38
rs771153816 Chr.3: 169796281-169796281 on Build GRCh38
rs902181414 Chr.3: 169796409-169796409 on Build GRCh38
rs767809406 Chr.3: 169796366-169796366 on Build GRCh38
rs143141600 Chr.3: 169796289-169796289 on Build GRCh38

View Full Product Details