Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154349316-154349561 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLNA
Entrez Gene ID
2316
Gene Name
filamin A
Gene Aliases
ABP-280, ABPX, CSBS, CVD1, FLN, FLN-A, FLN1, FMD, MNS, NHBP, OPD, OPD1, OPD2, XLVD, XMVD
UniGene
Hs.195464
Gene Chromosome Location
Chr.X: 154348529-154374638 on Build GRCh38
Transcripts
NM_001110556.1 NM_001456.3 XM_011531127.1 XM_011531128.1 XM_011531129.1 XM_011531130.1 XM_011531131.1

SNP Detail

SNP ID Chromosome Location
rs886039104 Chr.X: 154349514-154349514 on Build GRCh38
rs782006629 Chr.X: 154349403-154349403 on Build GRCh38
rs200694807 Chr.X: 154349522-154349522 on Build GRCh38
rs2070815 Chr.X: 154349441-154349441 on Build GRCh38
rs998077945 Chr.X: 154349506-154349506 on Build GRCh38
rs782784577 Chr.X: 154349450-154349450 on Build GRCh38
rs782291033 Chr.X: 154349404-154349404 on Build GRCh38
rs782594128 Chr.X: 154349355-154349355 on Build GRCh38
rs781867343 Chr.X: 154349343-154349343 on Build GRCh38
rs782375435 Chr.X: 154349393-154349393 on Build GRCh38
rs371724771 Chr.X: 154349469-154349469 on Build GRCh38
rs377518545 Chr.X: 154349380-154349380 on Build GRCh38
rs909656307 Chr.X: 154349459-154349459 on Build GRCh38
rs886039148 Chr.X: 154349510-154349510 on Build GRCh38
rs111243479 Chr.X: 154349360-154349360 on Build GRCh38
rs782263210 Chr.X: 154349504-154349504 on Build GRCh38
rs782496288 Chr.X: 154349491-154349491 on Build GRCh38
rs781930608 Chr.X: 154349406-154349406 on Build GRCh38
rs76337075 Chr.X: 154349432-154349432 on Build GRCh38
rs7063300 Chr.X: 154349351-154349351 on Build GRCh38
rs367666924 Chr.X: 154349462-154349462 on Build GRCh38
rs782651197 Chr.X: 154349496-154349496 on Build GRCh38
rs201762017 Chr.X: 154349490-154349490 on Build GRCh38
rs398123624 Chr.X: 154349447-154349447 on Build GRCh38
rs113471366 Chr.X: 154349359-154349359 on Build GRCh38
rs782535006 Chr.X: 154349345-154349345 on Build GRCh38
rs201883808 Chr.X: 154349534-154349534 on Build GRCh38
rs781983475 Chr.X: 154349435-154349435 on Build GRCh38
rs782686474 Chr.X: 154349350-154349350 on Build GRCh38
rs201663443 Chr.X: 154349354-154349354 on Build GRCh38
rs373872030 Chr.X: 154349451-154349451 on Build GRCh38
rs782308141 Chr.X: 154349405-154349405 on Build GRCh38
rs886044883 Chr.X: 154349506-154349506 on Build GRCh38
rs781905890 Chr.X: 154349468-154349468 on Build GRCh38
rs782714612 Chr.X: 154349488-154349488 on Build GRCh38
rs782125615 Chr.X: 154349449-154349449 on Build GRCh38
rs782709737 Chr.X: 154349408-154349408 on Build GRCh38

View Full Product Details