Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 150682181-150682418 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERICH6
Entrez Gene ID
131831
Gene Name
glutamate rich 6
Gene Aliases
C3orf44, ERICH6A, FAM194A
UniGene
Hs.147128
Gene Chromosome Location
Chr.3: 150659885-150704563 on Build GRCh38
Transcripts
NM_001308234.1 NM_152394.4 XM_005247120.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3588653 c.878G>A p.G293E Chr.3: 150682222-150682222 on Build GRCh38
3392121 c.856G>T p.D286Y Chr.3: 150682244-150682244 on Build GRCh38
5910366 c.863C>T p.S288F Chr.3: 150682237-150682237 on Build GRCh38
5799094 c.813T>C p.C271C Chr.3: 150682287-150682287 on Build GRCh38
1327804 c.856G>C p.D286H Chr.3: 150682244-150682244 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867827783 Chr.3: 150682369-150682369 on Build GRCh38
rs752326458 Chr.3: 150682323-150682323 on Build GRCh38
rs896763742 Chr.3: 150682384-150682384 on Build GRCh38
rs755644907 Chr.3: 150682241-150682241 on Build GRCh38
rs763281804 Chr.3: 150682299-150682299 on Build GRCh38
rs763608850 Chr.3: 150682361-150682361 on Build GRCh38
rs779069581 Chr.3: 150682342-150682344 on Build GRCh38
rs753857141 Chr.3: 150682368-150682368 on Build GRCh38
rs141068259 Chr.3: 150682387-150682387 on Build GRCh38
rs529540902 Chr.3: 150682261-150682261 on Build GRCh38
rs778807623 Chr.3: 150682245-150682245 on Build GRCh38
rs951017451 Chr.3: 150682391-150682391 on Build GRCh38
rs367591125 Chr.3: 150682266-150682266 on Build GRCh38
rs879169393 Chr.3: 150682390-150682390 on Build GRCh38
rs773608227 Chr.3: 150682281-150682281 on Build GRCh38
rs146020271 Chr.3: 150682348-150682349 on Build GRCh38
rs142979002 Chr.3: 150682243-150682243 on Build GRCh38
rs556312892 Chr.3: 150682271-150682271 on Build GRCh38
rs772084920 Chr.3: 150682356-150682360 on Build GRCh38
rs568717341 Chr.3: 150682248-150682248 on Build GRCh38
rs771166785 Chr.3: 150682302-150682304 on Build GRCh38
rs199606951 Chr.3: 150682272-150682272 on Build GRCh38
rs576508442 Chr.3: 150682217-150682217 on Build GRCh38
rs777576823 Chr.3: 150682242-150682242 on Build GRCh38
rs113746092 Chr.3: 150682301-150682301 on Build GRCh38
rs542080384 Chr.3: 150682312-150682312 on Build GRCh38
rs1002352445 Chr.3: 150682313-150682313 on Build GRCh38
rs767134111 Chr.3: 150682321-150682321 on Build GRCh38
rs1043801188 Chr.3: 150682290-150682290 on Build GRCh38
rs780306628 Chr.3: 150682270-150682270 on Build GRCh38
rs1006475386 Chr.3: 150682367-150682367 on Build GRCh38
rs1048996561 Chr.3: 150682320-150682320 on Build GRCh38

View Full Product Details