Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 24178221-24178482 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCDC2
Entrez Gene ID
51473
Gene Name
doublecortin domain containing 2
Gene Aliases
DCDC2A, DFNB66, NPHP19, RU2, RU2S
UniGene
Hs.61345
Gene Chromosome Location
Chr.6: 24171755-24383292 on Build GRCh38
Transcripts
NM_001195610.1 NM_016356.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4751309 c.1313G>C p.S438T Chr.6: 24178343-24178343 on Build GRCh38
6326489 c.454G>A p.A152T Chr.6: 24178461-24178461 on Build GRCh38
6477084 c.575G>T p.G192V Chr.6: 24178340-24178340 on Build GRCh38
7109410 c.1233G>T p.E411D Chr.6: 24178423-24178423 on Build GRCh38
6483189 c.443G>T p.S148I Chr.6: 24178472-24178472 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754991150 Chr.6: 24178444-24178444 on Build GRCh38
rs756047736 Chr.6: 24178437-24178437 on Build GRCh38
rs749297858 Chr.6: 24178383-24178383 on Build GRCh38
rs794729665 Chr.6: 24178385-24178385 on Build GRCh38
rs756984644 Chr.6: 24178365-24178365 on Build GRCh38
rs201830443 Chr.6: 24178342-24178342 on Build GRCh38
rs549447263 Chr.6: 24178442-24178442 on Build GRCh38
rs752452396 Chr.6: 24178321-24178321 on Build GRCh38
rs773832570 Chr.6: 24178415-24178415 on Build GRCh38
rs370330889 Chr.6: 24178251-24178254 on Build GRCh38
rs1010694033 Chr.6: 24178292-24178292 on Build GRCh38
rs547134434 Chr.6: 24178417-24178417 on Build GRCh38
rs567036123 Chr.6: 24178435-24178435 on Build GRCh38
rs530744307 Chr.6: 24178270-24178270 on Build GRCh38
rs878958751 Chr.6: 24178423-24178423 on Build GRCh38
rs758852015 Chr.6: 24178452-24178452 on Build GRCh38
rs751396384 Chr.6: 24178338-24178338 on Build GRCh38
rs1052123949 Chr.6: 24178273-24178273 on Build GRCh38
rs78242664 Chr.6: 24178271-24178271 on Build GRCh38
rs763616532 Chr.6: 24178336-24178336 on Build GRCh38
rs759153113 Chr.6: 24178302-24178302 on Build GRCh38
rs763861048 Chr.6: 24178432-24178432 on Build GRCh38
rs771499861 Chr.6: 24178419-24178419 on Build GRCh38
rs777259245 Chr.6: 24178458-24178458 on Build GRCh38
rs139858268 Chr.6: 24178448-24178448 on Build GRCh38
rs747339767 Chr.6: 24178454-24178454 on Build GRCh38
rs771591530 Chr.6: 24178457-24178457 on Build GRCh38
rs748310193 Chr.6: 24178451-24178451 on Build GRCh38
rs772289717 Chr.6: 24178403-24178403 on Build GRCh38
rs748591010 Chr.6: 24178401-24178401 on Build GRCh38
rs138279131 Chr.6: 24178344-24178344 on Build GRCh38
rs778953600 Chr.6: 24178393-24178393 on Build GRCh38
rs755258271 Chr.6: 24178384-24178384 on Build GRCh38
rs375119774 Chr.6: 24178373-24178373 on Build GRCh38
rs149268081 Chr.6: 24178436-24178436 on Build GRCh38
rs531483232 Chr.6: 24178249-24178252 on Build GRCh38
rs143452599 Chr.6: 24178453-24178453 on Build GRCh38
rs929868382 Chr.6: 24178262-24178262 on Build GRCh38
rs561132766 Chr.6: 24178311-24178311 on Build GRCh38
rs960014369 Chr.6: 24178391-24178391 on Build GRCh38
rs530158986 Chr.6: 24178324-24178324 on Build GRCh38
rs762042617 Chr.6: 24178286-24178286 on Build GRCh38
rs781230955 Chr.6: 24178368-24178368 on Build GRCh38
rs750357959 Chr.6: 24178372-24178372 on Build GRCh38
rs772655581 Chr.6: 24178296-24178296 on Build GRCh38
rs775174265 Chr.6: 24178431-24178431 on Build GRCh38
rs143521294 Chr.6: 24178443-24178443 on Build GRCh38
rs551374870 Chr.6: 24178407-24178407 on Build GRCh38
rs553676240 Chr.6: 24178413-24178413 on Build GRCh38
rs534996877 Chr.6: 24178381-24178381 on Build GRCh38
rs773566102 Chr.6: 24178298-24178298 on Build GRCh38

View Full Product Details