Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 48963219-48963470 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSHR
Entrez Gene ID
2492
Gene Name
follicle stimulating hormone receptor
Gene Aliases
FSHR1, FSHRO, LGR1, ODG1
UniGene
Hs.1428
Gene Chromosome Location
Chr.2: 48953161-49154527 on Build GRCh38
Transcripts
NM_000145.3 NM_181446.2 XM_011532733.2 XM_011532734.2 XM_011532735.2 XM_011532736.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3391750 c.1531T>C p.Y511H Chr.2: 48963290-48963290 on Build GRCh38
721654 c.1482G>C p.W494C Chr.2: 48963339-48963339 on Build GRCh38
265445 c.1415C>A p.T472K Chr.2: 48963406-48963406 on Build GRCh38
5719593 c.1440G>A p.K480K Chr.2: 48963381-48963381 on Build GRCh38
721655 c.1527C>A p.S509R Chr.2: 48963294-48963294 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781192232 Chr.2: 48963360-48963360 on Build GRCh38
rs576761459 Chr.2: 48963405-48963405 on Build GRCh38
rs756512445 Chr.2: 48963317-48963317 on Build GRCh38
rs889044499 Chr.2: 48963434-48963434 on Build GRCh38
rs866359351 Chr.2: 48963350-48963350 on Build GRCh38
rs763241241 Chr.2: 48963370-48963370 on Build GRCh38
rs150549219 Chr.2: 48963346-48963346 on Build GRCh38
rs780625359 Chr.2: 48963439-48963439 on Build GRCh38
rs762018429 Chr.2: 48963380-48963380 on Build GRCh38
rs780337084 Chr.2: 48963337-48963337 on Build GRCh38
rs759405654 Chr.2: 48963261-48963261 on Build GRCh38
rs756806040 Chr.2: 48963438-48963438 on Build GRCh38
rs774526533 Chr.2: 48963375-48963375 on Build GRCh38
rs267599403 Chr.2: 48963241-48963241 on Build GRCh38
rs751002380 Chr.2: 48963421-48963421 on Build GRCh38
rs764264620 Chr.2: 48963289-48963289 on Build GRCh38
rs141624278 Chr.2: 48963363-48963363 on Build GRCh38
rs766665626 Chr.2: 48963343-48963343 on Build GRCh38
rs763325688 Chr.2: 48963277-48963277 on Build GRCh38
rs775795358 Chr.2: 48963276-48963276 on Build GRCh38
rs750925845 Chr.2: 48963391-48963391 on Build GRCh38
rs751733216 Chr.2: 48963297-48963297 on Build GRCh38
rs777241968 Chr.2: 48963343-48963343 on Build GRCh38
rs750715847 Chr.2: 48963307-48963307 on Build GRCh38
rs151318189 Chr.2: 48963256-48963256 on Build GRCh38
rs758082845 Chr.2: 48963339-48963339 on Build GRCh38
rs765311964 Chr.2: 48963267-48963267 on Build GRCh38
rs6167 Chr.2: 48963249-48963249 on Build GRCh38
rs368888734 Chr.2: 48963390-48963390 on Build GRCh38
rs114528248 Chr.2: 48963411-48963411 on Build GRCh38
rs121909662 Chr.2: 48963266-48963266 on Build GRCh38
rs769052497 Chr.2: 48963371-48963371 on Build GRCh38
rs770813496 Chr.2: 48963355-48963355 on Build GRCh38
rs746917603 Chr.2: 48963252-48963252 on Build GRCh38
rs775216471 Chr.2: 48963367-48963367 on Build GRCh38
rs932237074 Chr.2: 48963400-48963400 on Build GRCh38
rs777434194 Chr.2: 48963418-48963418 on Build GRCh38
rs770903417 Chr.2: 48963253-48963253 on Build GRCh38
rs762921549 Chr.2: 48963309-48963309 on Build GRCh38
rs752385959 Chr.2: 48963338-48963338 on Build GRCh38
rs935899649 Chr.2: 48963396-48963396 on Build GRCh38
rs745593936 Chr.2: 48963362-48963362 on Build GRCh38
rs138281715 Chr.2: 48963245-48963245 on Build GRCh38
rs766731004 Chr.2: 48963406-48963406 on Build GRCh38

View Full Product Details