Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 196672162-196672417 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC150
Entrez Gene ID
284992
Gene Name
coiled-coil domain containing 150
Gene Aliases
-
UniGene
Hs.132519
Gene Chromosome Location
Chr.2: 196639632-196733668 on Build GRCh38
Transcripts
NM_001080539.1 XM_006712438.3 XM_011510985.2 XM_011510987.2 XM_011510988.2 XM_017003861.1 XM_017003862.1 XM_017003863.1 XM_017003864.1 XM_017003865.1 XM_017003866.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1631697 c.971A>G p.E324G Chr.2: 196672379-196672379 on Build GRCh38
1014408 c.942G>A p.L314L Chr.2: 196672350-196672350 on Build GRCh38
6888586 c.942G>T p.L314L Chr.2: 196672350-196672350 on Build GRCh38
6010584 c.1006C>T p.H336Y Chr.2: 196672414-196672414 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs757880806 Chr.2: 196672372-196672372 on Build GRCh38
rs534755954 Chr.2: 196672342-196672342 on Build GRCh38
rs765516875 Chr.2: 196672325-196672325 on Build GRCh38
rs190816467 Chr.2: 196672254-196672254 on Build GRCh38
rs781760040 Chr.2: 196672373-196672373 on Build GRCh38
rs889488703 Chr.2: 196672371-196672371 on Build GRCh38
rs16856309 Chr.2: 196672291-196672291 on Build GRCh38
rs547672187 Chr.2: 196672386-196672386 on Build GRCh38
rs868677733 Chr.2: 196672369-196672369 on Build GRCh38
rs959391811 Chr.2: 196672289-196672289 on Build GRCh38
rs183860842 Chr.2: 196672263-196672263 on Build GRCh38
rs76753956 Chr.2: 196672338-196672338 on Build GRCh38
rs754732344 Chr.2: 196672301-196672301 on Build GRCh38
rs778058598 Chr.2: 196672345-196672345 on Build GRCh38
rs187165523 Chr.2: 196672271-196672271 on Build GRCh38
rs993272365 Chr.2: 196672219-196672219 on Build GRCh38
rs529923074 Chr.2: 196672197-196672197 on Build GRCh38
rs143988285 Chr.2: 196672190-196672190 on Build GRCh38
rs148697217 Chr.2: 196672391-196672391 on Build GRCh38
rs751967908 Chr.2: 196672363-196672363 on Build GRCh38
rs567936623 Chr.2: 196672255-196672255 on Build GRCh38
rs752820765 Chr.2: 196672330-196672330 on Build GRCh38
rs769816304 Chr.2: 196672196-196672196 on Build GRCh38
rs748831818 Chr.2: 196672377-196672377 on Build GRCh38
rs963218916 Chr.2: 196672201-196672201 on Build GRCh38
rs745699093 Chr.2: 196672240-196672240 on Build GRCh38
rs926270310 Chr.2: 196672286-196672286 on Build GRCh38
rs771987095 Chr.2: 196672329-196672330 on Build GRCh38
rs780168007 Chr.2: 196672330-196672330 on Build GRCh38
rs753791580 Chr.2: 196672217-196672217 on Build GRCh38

View Full Product Details