Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6730493-6730744 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPS7A
Entrez Gene ID
50813
Gene Name
COP9 signalosome subunit 7A
Gene Aliases
CSN7, CSN7A, SGN7a
UniGene
Hs.530823
Gene Chromosome Location
Chr.12: 6723984-6731875 on Build GRCh38
Transcripts
NM_001164093.1 NM_001164094.1 NM_001164095.2 NM_016319.3 XM_005253694.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7250804 c.697C>G p.Q233E Chr.12: 6730729-6730729 on Build GRCh38
7201094 c.658A>G p.I220V Chr.12: 6730690-6730690 on Build GRCh38
1363860 c.675A>G p.A225A Chr.12: 6730707-6730707 on Build GRCh38
6634106 c.656C>A p.T219N Chr.12: 6730688-6730688 on Build GRCh38
4044407 c.665T>G p.V222G Chr.12: 6730697-6730697 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771057632 Chr.12: 6730518-6730519 on Build GRCh38
rs779854611 Chr.12: 6730518-6730518 on Build GRCh38
rs549562987 Chr.12: 6730723-6730723 on Build GRCh38
rs770530289 Chr.12: 6730637-6730637 on Build GRCh38
rs146849329 Chr.12: 6730665-6730665 on Build GRCh38
rs1010286935 Chr.12: 6730707-6730707 on Build GRCh38
rs986850184 Chr.12: 6730580-6730580 on Build GRCh38
rs545720525 Chr.12: 6730524-6730524 on Build GRCh38
rs1054593291 Chr.12: 6730699-6730699 on Build GRCh38
rs929949885 Chr.12: 6730575-6730575 on Build GRCh38
rs199950835 Chr.12: 6730716-6730716 on Build GRCh38
rs758548997 Chr.12: 6730517-6730517 on Build GRCh38
rs754699471 Chr.12: 6730525-6730525 on Build GRCh38
rs565116686 Chr.12: 6730654-6730654 on Build GRCh38
rs766481814 Chr.12: 6730704-6730704 on Build GRCh38
rs762321124 Chr.12: 6730659-6730659 on Build GRCh38
rs748743191 Chr.12: 6730625-6730625 on Build GRCh38
rs780993893 Chr.12: 6730542-6730542 on Build GRCh38
rs942951678 Chr.12: 6730618-6730618 on Build GRCh38
rs758397165 Chr.12: 6730703-6730703 on Build GRCh38
rs747746777 Chr.12: 6730554-6730554 on Build GRCh38
rs769410657 Chr.12: 6730555-6730555 on Build GRCh38
rs369598803 Chr.12: 6730624-6730624 on Build GRCh38
rs374957064 Chr.12: 6730647-6730647 on Build GRCh38
rs1039858688 Chr.12: 6730627-6730627 on Build GRCh38
rs912778956 Chr.12: 6730600-6730600 on Build GRCh38
rs774230583 Chr.12: 6730698-6730699 on Build GRCh38
rs750727471 Chr.12: 6730701-6730701 on Build GRCh38
rs200625984 Chr.12: 6730642-6730642 on Build GRCh38
rs148675442 Chr.12: 6730717-6730717 on Build GRCh38

View Full Product Details