Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118979004-118979271 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXR1
Entrez Gene ID
283150
Gene Name
forkhead box R1
Gene Aliases
DLNB13, FOXN5
UniGene
Hs.116679
Gene Chromosome Location
Chr.11: 118971398-118981601 on Build GRCh38
Transcripts
NM_181721.2 XM_017017575.1 XM_017017576.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7368690 c.190C>G p.P64A Chr.11: 118979010-118979010 on Build GRCh38
6461571 c.346C>T p.P116S Chr.11: 118979166-118979166 on Build GRCh38
7368692 c.380G>T p.W127L Chr.11: 118979200-118979200 on Build GRCh38
4988355 c.225G>C p.K75N Chr.11: 118979045-118979045 on Build GRCh38
1162385 c.310G>A p.E104K Chr.11: 118979130-118979130 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782667971 Chr.11: 118979183-118979183 on Build GRCh38
rs572984762 Chr.11: 118979164-118979164 on Build GRCh38
rs782252134 Chr.11: 118979177-118979177 on Build GRCh38
rs782419728 Chr.11: 118979135-118979135 on Build GRCh38
rs80305910 Chr.11: 118979037-118979037 on Build GRCh38
rs375614430 Chr.11: 118979039-118979039 on Build GRCh38
rs183794856 Chr.11: 118979178-118979178 on Build GRCh38
rs782458567 Chr.11: 118979045-118979045 on Build GRCh38
rs782213316 Chr.11: 118979229-118979229 on Build GRCh38
rs781855866 Chr.11: 118979063-118979063 on Build GRCh38
rs782511902 Chr.11: 118979064-118979064 on Build GRCh38
rs782479317 Chr.11: 118979175-118979175 on Build GRCh38
rs200679607 Chr.11: 118979201-118979201 on Build GRCh38
rs782112617 Chr.11: 118979159-118979159 on Build GRCh38
rs149060377 Chr.11: 118979088-118979088 on Build GRCh38
rs781872934 Chr.11: 118979028-118979028 on Build GRCh38
rs963712326 Chr.11: 118979071-118979071 on Build GRCh38
rs143117752 Chr.11: 118979121-118979121 on Build GRCh38
rs77188008 Chr.11: 118979120-118979120 on Build GRCh38
rs781802658 Chr.11: 118979173-118979173 on Build GRCh38
rs782612307 Chr.11: 118979176-118979176 on Build GRCh38
rs782134663 Chr.11: 118979125-118979125 on Build GRCh38
rs782052870 Chr.11: 118979051-118979051 on Build GRCh38
rs61902072 Chr.11: 118979129-118979129 on Build GRCh38
rs782374108 Chr.11: 118979233-118979233 on Build GRCh38
rs782771981 Chr.11: 118979167-118979167 on Build GRCh38
rs1024339047 Chr.11: 118979247-118979247 on Build GRCh38
rs545792564 Chr.11: 118979068-118979068 on Build GRCh38
rs112326280 Chr.11: 118979246-118979246 on Build GRCh38
rs887029608 Chr.11: 118979033-118979033 on Build GRCh38
rs144422939 Chr.11: 118979170-118979170 on Build GRCh38
rs901491412 Chr.11: 118979087-118979087 on Build GRCh38
rs376486817 Chr.11: 118979074-118979074 on Build GRCh38
rs782541995 Chr.11: 118979169-118979169 on Build GRCh38
rs967638864 Chr.11: 118979205-118979205 on Build GRCh38
rs560972749 Chr.11: 118979158-118979158 on Build GRCh38
rs781944088 Chr.11: 118979213-118979213 on Build GRCh38
rs782038004 Chr.11: 118979136-118979136 on Build GRCh38
rs782591291 Chr.11: 118979086-118979086 on Build GRCh38
rs782306764 Chr.11: 118979076-118979076 on Build GRCh38
rs782313873 Chr.11: 118979209-118979209 on Build GRCh38
rs782613972 Chr.11: 118979046-118979046 on Build GRCh38
rs782754029 Chr.11: 118979026-118979026 on Build GRCh38
rs781992140 Chr.11: 118979238-118979238 on Build GRCh38
rs782821168 Chr.11: 118979038-118979038 on Build GRCh38
rs782055229 Chr.11: 118979157-118979157 on Build GRCh38

View Full Product Details