Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89180277-89180520 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH15
Entrez Gene ID
1013
Gene Name
cadherin 15
Gene Aliases
CDH14, CDH3, CDHM, MCAD, MRD3
UniGene
Hs.148090
Gene Chromosome Location
Chr.16: 89171755-89195492 on Build GRCh38
Transcripts
NM_004933.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5683112 c.280G>T p.V94F Chr.16: 89180278-89180278 on Build GRCh38
2922802 c.334C>T p.R112C Chr.16: 89180332-89180332 on Build GRCh38
3513253 c.285C>T p.F95F Chr.16: 89180283-89180283 on Build GRCh38
1200424 c.335G>A p.R112H Chr.16: 89180333-89180333 on Build GRCh38
2922803 c.336C>T p.R112R Chr.16: 89180334-89180334 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142654313 Chr.16: 89180479-89180479 on Build GRCh38
rs547157882 Chr.16: 89180443-89180443 on Build GRCh38
rs923171052 Chr.16: 89180331-89180331 on Build GRCh38
rs144975891 Chr.16: 89180318-89180318 on Build GRCh38
rs758676906 Chr.16: 89180321-89180321 on Build GRCh38
rs201228695 Chr.16: 89180332-89180332 on Build GRCh38
rs200552002 Chr.16: 89180367-89180367 on Build GRCh38
rs377464592 Chr.16: 89180379-89180379 on Build GRCh38
rs1038051720 Chr.16: 89180485-89180485 on Build GRCh38
rs374291126 Chr.16: 89180312-89180312 on Build GRCh38
rs1018449179 Chr.16: 89180364-89180364 on Build GRCh38
rs768597663 Chr.16: 89180349-89180349 on Build GRCh38
rs760631737 Chr.16: 89180305-89180305 on Build GRCh38
rs548474638 Chr.16: 89180484-89180484 on Build GRCh38
rs543467772 Chr.16: 89180322-89180322 on Build GRCh38
rs765376733 Chr.16: 89180317-89180317 on Build GRCh38
rs993248194 Chr.16: 89180340-89180340 on Build GRCh38
rs546580091 Chr.16: 89180421-89180421 on Build GRCh38
rs113385148 Chr.16: 89180309-89180309 on Build GRCh38
rs532278777 Chr.16: 89180433-89180433 on Build GRCh38
rs371999213 Chr.16: 89180373-89180373 on Build GRCh38
rs756755137 Chr.16: 89180378-89180378 on Build GRCh38
rs578055365 Chr.16: 89180347-89180347 on Build GRCh38
rs374302975 Chr.16: 89180334-89180334 on Build GRCh38
rs147758898 Chr.16: 89180360-89180360 on Build GRCh38
rs767586241 Chr.16: 89180374-89180374 on Build GRCh38
rs564789076 Chr.16: 89180426-89180426 on Build GRCh38
rs756201732 Chr.16: 89180390-89180390 on Build GRCh38
rs149088017 Chr.16: 89180333-89180333 on Build GRCh38
rs569104731 Chr.16: 89180448-89180448 on Build GRCh38
rs761821061 Chr.16: 89180356-89180356 on Build GRCh38
rs765286825 Chr.16: 89180359-89180359 on Build GRCh38
rs776437019 Chr.16: 89180350-89180350 on Build GRCh38
rs116889318 Chr.16: 89180377-89180377 on Build GRCh38
rs913203489 Chr.16: 89180357-89180357 on Build GRCh38
rs369815248 Chr.16: 89180445-89180445 on Build GRCh38
rs778535812 Chr.16: 89180385-89180385 on Build GRCh38
rs756843357 Chr.16: 89180335-89180335 on Build GRCh38
rs115218393 Chr.16: 89180403-89180403 on Build GRCh38
rs754343628 Chr.16: 89180498-89180498 on Build GRCh38
rs1005809887 Chr.16: 89180468-89180468 on Build GRCh38
rs368121374 Chr.16: 89180368-89180368 on Build GRCh38
rs148904837 Chr.16: 89180447-89180447 on Build GRCh38
rs745421861 Chr.16: 89180391-89180391 on Build GRCh38
rs763987083 Chr.16: 89180310-89180310 on Build GRCh38
rs758075235 Chr.16: 89180394-89180394 on Build GRCh38
rs761910970 Chr.16: 89180316-89180316 on Build GRCh38
rs745513122 Chr.16: 89180344-89180344 on Build GRCh38
rs746751282 Chr.16: 89180395-89180395 on Build GRCh38
rs746803500 Chr.16: 89180348-89180348 on Build GRCh38

View Full Product Details