Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 81225303-81225576 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL16
Entrez Gene ID
3603
Gene Name
interleukin 16
Gene Aliases
LCF, NIL16, PRIL16, prIL-16
UniGene
Hs.459095
Gene Chromosome Location
Chr.15: 81181844-81312763 on Build GRCh38
Transcripts
NM_001172128.1 NM_172217.3 XM_005254342.3 XM_011521519.2 XM_011521520.2 XM_017022140.1 XM_017022141.1 XM_017022142.1 XM_017022143.1 XM_017022144.1 XM_017022145.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7322965 c.112G>A p.D38N Chr.15: 81225511-81225511 on Build GRCh38
5173717 c.18C>T p.R6R Chr.15: 81225417-81225417 on Build GRCh38
5754738 c.153G>T p.Q51H Chr.15: 81225552-81225552 on Build GRCh38
4878095 c.14G>T p.S5I Chr.15: 81225413-81225413 on Build GRCh38
331796 c.131C>A p.P44H Chr.15: 81225530-81225530 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778733686 Chr.15: 81225429-81225429 on Build GRCh38
rs191307785 Chr.15: 81225416-81225416 on Build GRCh38
rs773559808 Chr.15: 81225367-81225367 on Build GRCh38
rs376483266 Chr.15: 81225341-81225341 on Build GRCh38
rs776705538 Chr.15: 81225402-81225402 on Build GRCh38
rs750669821 Chr.15: 81225444-81225444 on Build GRCh38
rs760626201 Chr.15: 81225530-81225530 on Build GRCh38
rs925368936 Chr.15: 81225360-81225360 on Build GRCh38
rs775546068 Chr.15: 81225468-81225468 on Build GRCh38
rs777933017 Chr.15: 81225553-81225553 on Build GRCh38
rs773073585 Chr.15: 81225506-81225506 on Build GRCh38
rs759686946 Chr.15: 81225407-81225407 on Build GRCh38
rs750324638 Chr.15: 81225418-81225418 on Build GRCh38
rs764866226 Chr.15: 81225550-81225550 on Build GRCh38
rs200683061 Chr.15: 81225340-81225340 on Build GRCh38
rs183847813 Chr.15: 81225431-81225431 on Build GRCh38
rs576209749 Chr.15: 81225417-81225417 on Build GRCh38
rs745968376 Chr.15: 81225450-81225450 on Build GRCh38
rs758682758 Chr.15: 81225432-81225432 on Build GRCh38
rs924078060 Chr.15: 81225531-81225531 on Build GRCh38
rs747412827 Chr.15: 81225440-81225440 on Build GRCh38
rs779357406 Chr.15: 81225338-81225338 on Build GRCh38
rs776652388 Chr.15: 81225449-81225449 on Build GRCh38
rs145164115 Chr.15: 81225465-81225465 on Build GRCh38
rs759598447 Chr.15: 81225393-81225393 on Build GRCh38
rs756134523 Chr.15: 81225422-81225422 on Build GRCh38
rs753406289 Chr.15: 81225426-81225426 on Build GRCh38
rs565192328 Chr.15: 81225435-81225435 on Build GRCh38
rs767203646 Chr.15: 81225503-81225503 on Build GRCh38
rs770980958 Chr.15: 81225369-81225369 on Build GRCh38
rs753351248 Chr.15: 81225545-81225545 on Build GRCh38
rs373015962 Chr.15: 81225494-81225494 on Build GRCh38
rs752276629 Chr.15: 81225551-81225551 on Build GRCh38
rs754641043 Chr.15: 81225427-81225427 on Build GRCh38
rs772584232 Chr.15: 81225347-81225347 on Build GRCh38
rs765933501 Chr.15: 81225425-81225425 on Build GRCh38
rs955553429 Chr.15: 81225486-81225486 on Build GRCh38
rs202011202 Chr.15: 81225415-81225415 on Build GRCh38
rs758078437 Chr.15: 81225552-81225552 on Build GRCh38
rs766454583 Chr.15: 81225541-81225541 on Build GRCh38
rs968249097 Chr.15: 81225493-81225493 on Build GRCh38
rs979572700 Chr.15: 81225497-81225497 on Build GRCh38
rs761308358 Chr.15: 81225368-81225368 on Build GRCh38
rs201225720 Chr.15: 81225408-81225408 on Build GRCh38
rs199973594 Chr.15: 81225448-81225448 on Build GRCh38

View Full Product Details