Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 32462498-32462747 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC7
Entrez Gene ID
79741
Gene Name
coiled-coil domain containing 7
Gene Aliases
BIOT2, BioT2-A, BioT2-B, BioT2-C, C10orf68
UniGene
Hs.585464
Gene Chromosome Location
Chr.10: 32446082-32882864 on Build GRCh38
Transcripts
NM_001026383.2 NM_001321115.1 NM_145023.5 NR_109826.1 NR_109827.1 XM_006717505.1 XM_006717506.1 XM_011519663.1 XM_011519664.1 XM_011519666.1 XM_011519667.1 XM_011519668.1 XM_011519669.1 XM_011519670.1 XM_011519671.1 XM_011519672.1 XM_011519673.1 XM_011519674.1 XM_011519675.1 XM_011519676.1 XM_011519677.1 XM_011519678.1 XM_011519679.1 XM_011519684.2 XM_011519686.2 XM_011519688.2 XM_017016638.1 XM_017016639.1 XM_017016640.1 XM_017016643.1 XM_017016644.1 XM_017016645.1 XM_017016647.1 XM_017016650.1 XM_017016651.1

SNP Detail

SNP ID Chromosome Location
rs780429855 Chr.10: 32462641-32462641 on Build GRCh38
rs770407353 Chr.10: 32462708-32462708 on Build GRCh38
rs189950131 Chr.10: 32462578-32462578 on Build GRCh38
rs530682765 Chr.10: 32462689-32462689 on Build GRCh38
rs758738610 Chr.10: 32462634-32462634 on Build GRCh38
rs755804541 Chr.10: 32462666-32462666 on Build GRCh38
rs892121335 Chr.10: 32462717-32462717 on Build GRCh38
rs900626378 Chr.10: 32462533-32462533 on Build GRCh38
rs564374932 Chr.10: 32462564-32462564 on Build GRCh38
rs777279863 Chr.10: 32462618-32462622 on Build GRCh38
rs1050556776 Chr.10: 32462657-32462657 on Build GRCh38
rs748830302 Chr.10: 32462707-32462707 on Build GRCh38
rs923478728 Chr.10: 32462623-32462623 on Build GRCh38
rs546198006 Chr.10: 32462537-32462537 on Build GRCh38
rs910334546 Chr.10: 32462663-32462663 on Build GRCh38
rs771951175 Chr.10: 32462715-32462715 on Build GRCh38
rs777084739 Chr.10: 32462703-32462703 on Build GRCh38
rs199581902 Chr.10: 32462656-32462656 on Build GRCh38
rs774411770 Chr.10: 32462712-32462712 on Build GRCh38
rs771603962 Chr.10: 32462674-32462674 on Build GRCh38
rs745838102 Chr.10: 32462713-32462713 on Build GRCh38
rs1028123001 Chr.10: 32462597-32462597 on Build GRCh38
rs12416437 Chr.10: 32462598-32462598 on Build GRCh38
rs182939095 Chr.10: 32462654-32462654 on Build GRCh38
rs909213660 Chr.10: 32462531-32462531 on Build GRCh38
rs761324106 Chr.10: 32462616-32462616 on Build GRCh38

View Full Product Details