Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 8066082-8066315 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBN3
Entrez Gene ID
84467
Gene Name
fibrillin 3
Gene Aliases
-
UniGene
Hs.370362
Gene Chromosome Location
Chr.19: 8065402-8149846 on Build GRCh38
Transcripts
NM_001321431.1 NM_032447.4 XM_017027372.1 XM_017027373.1 XM_017027374.1 XM_017027375.1 XM_017027376.1 XM_017027377.1 XM_017027378.1 XM_017027379.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1252223 c.8246T>A p.F2749Y Chr.19: 8066103-8066103 on Build GRCh38
6046494 c.8153G>A p.G2718D Chr.19: 8066196-8066196 on Build GRCh38
5694179 c.8159C>A p.A2720D Chr.19: 8066190-8066190 on Build GRCh38
3541503 c.8223C>T p.I2741I Chr.19: 8066126-8066126 on Build GRCh38
3693337 c.8161G>T p.E2721* Chr.19: 8066188-8066188 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139990648 Chr.19: 8066219-8066219 on Build GRCh38
rs756201240 Chr.19: 8066137-8066137 on Build GRCh38
rs779357354 Chr.19: 8066106-8066106 on Build GRCh38
rs755536126 Chr.19: 8066201-8066201 on Build GRCh38
rs747732485 Chr.19: 8066124-8066124 on Build GRCh38
rs368561585 Chr.19: 8066131-8066131 on Build GRCh38
rs780834583 Chr.19: 8066120-8066120 on Build GRCh38
rs780445413 Chr.19: 8066164-8066164 on Build GRCh38
rs370743995 Chr.19: 8066250-8066250 on Build GRCh38
rs746542118 Chr.19: 8066114-8066114 on Build GRCh38
rs917658642 Chr.19: 8066121-8066121 on Build GRCh38
rs148868478 Chr.19: 8066142-8066142 on Build GRCh38
rs752241003 Chr.19: 8066214-8066214 on Build GRCh38
rs375861193 Chr.19: 8066191-8066191 on Build GRCh38
rs955076879 Chr.19: 8066263-8066263 on Build GRCh38
rs901264220 Chr.19: 8066174-8066174 on Build GRCh38
rs34941422 Chr.19: 8066119-8066119 on Build GRCh38
rs758760112 Chr.19: 8066161-8066161 on Build GRCh38
rs148313867 Chr.19: 8066150-8066150 on Build GRCh38
rs770712458 Chr.19: 8066173-8066173 on Build GRCh38
rs545168335 Chr.19: 8066184-8066184 on Build GRCh38
rs765545923 Chr.19: 8066107-8066108 on Build GRCh38
rs144078033 Chr.19: 8066193-8066193 on Build GRCh38
rs750803710 Chr.19: 8066155-8066155 on Build GRCh38
rs75174438 Chr.19: 8066169-8066169 on Build GRCh38
rs150683901 Chr.19: 8066194-8066194 on Build GRCh38
rs539996236 Chr.19: 8066282-8066282 on Build GRCh38
rs748916084 Chr.19: 8066225-8066225 on Build GRCh38
rs558440722 Chr.19: 8066188-8066188 on Build GRCh38
rs745645966 Chr.19: 8066255-8066255 on Build GRCh38
rs143150244 Chr.19: 8066166-8066166 on Build GRCh38
rs374699801 Chr.19: 8066167-8066167 on Build GRCh38
rs138322146 Chr.19: 8066189-8066189 on Build GRCh38
rs199602063 Chr.19: 8066170-8066170 on Build GRCh38
rs758887259 Chr.19: 8066109-8066109 on Build GRCh38
rs760693740 Chr.19: 8066136-8066136 on Build GRCh38
rs372313415 Chr.19: 8066125-8066125 on Build GRCh38
rs775538659 Chr.19: 8066290-8066290 on Build GRCh38
rs967738909 Chr.19: 8066237-8066237 on Build GRCh38
rs780590806 Chr.19: 8066113-8066113 on Build GRCh38
rs544478907 Chr.19: 8066203-8066203 on Build GRCh38
rs762783717 Chr.19: 8066128-8066128 on Build GRCh38
rs200637796 Chr.19: 8066185-8066185 on Build GRCh38
rs752046207 Chr.19: 8066165-8066165 on Build GRCh38
rs757011216 Chr.19: 8066236-8066236 on Build GRCh38
rs777315852 Chr.19: 8066221-8066221 on Build GRCh38
rs374334624 Chr.19: 8066126-8066126 on Build GRCh38
rs143551340 Chr.19: 8066143-8066143 on Build GRCh38
rs774171420 Chr.19: 8066180-8066180 on Build GRCh38
rs770811627 Chr.19: 8066132-8066132 on Build GRCh38
rs371800341 Chr.19: 8066137-8066137 on Build GRCh38
rs571628348 Chr.19: 8066107-8066107 on Build GRCh38

View Full Product Details