Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 30289145-30289400 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRTAP25-1
Entrez Gene ID
100131902
Gene Name
keratin associated protein 25-1
Gene Aliases
KAP25.1
UniGene
Hs.580882
Gene Chromosome Location
Chr.21: 30289145-30289514 on Build GRCh38
Transcripts
NM_001128598.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5690575 c.190A>T p.R64* Chr.21: 30289301-30289301 on Build GRCh38
1029864 c.173C>A p.S58Y Chr.21: 30289318-30289318 on Build GRCh38
3363411 c.166A>G p.I56V Chr.21: 30289325-30289325 on Build GRCh38
3758825 c.156T>C p.G52G Chr.21: 30289335-30289335 on Build GRCh38
3423894 c.299G>T p.R100I Chr.21: 30289192-30289192 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756783601 Chr.21: 30289301-30289301 on Build GRCh38
rs762639203 Chr.21: 30289201-30289201 on Build GRCh38
rs529648689 Chr.21: 30289348-30289348 on Build GRCh38
rs780351583 Chr.21: 30289352-30289352 on Build GRCh38
rs371589456 Chr.21: 30289321-30289321 on Build GRCh38
rs938905227 Chr.21: 30289315-30289315 on Build GRCh38
rs189774872 Chr.21: 30289314-30289314 on Build GRCh38
rs921917027 Chr.21: 30289251-30289251 on Build GRCh38
rs569441448 Chr.21: 30289168-30289168 on Build GRCh38
rs2832759 Chr.21: 30289335-30289335 on Build GRCh38
rs746746223 Chr.21: 30289362-30289362 on Build GRCh38
rs986055481 Chr.21: 30289211-30289211 on Build GRCh38
rs761142508 Chr.21: 30289266-30289266 on Build GRCh38
rs772855389 Chr.21: 30289185-30289185 on Build GRCh38
rs376985630 Chr.21: 30289275-30289275 on Build GRCh38
rs8127420 Chr.21: 30289373-30289373 on Build GRCh38
rs908169881 Chr.21: 30289293-30289293 on Build GRCh38
rs770295087 Chr.21: 30289313-30289313 on Build GRCh38
rs546401866 Chr.21: 30289193-30289193 on Build GRCh38
rs752965566 Chr.21: 30289237-30289237 on Build GRCh38
rs750036274 Chr.21: 30289358-30289358 on Build GRCh38
rs558036599 Chr.21: 30289216-30289216 on Build GRCh38
rs1044181521 Chr.21: 30289337-30289337 on Build GRCh38
rs918101508 Chr.21: 30289323-30289323 on Build GRCh38
rs774100936 Chr.21: 30289332-30289337 on Build GRCh38
rs145146360 Chr.21: 30289288-30289288 on Build GRCh38
rs942081236 Chr.21: 30289217-30289217 on Build GRCh38
rs952105392 Chr.21: 30289279-30289279 on Build GRCh38
rs940953195 Chr.21: 30289317-30289317 on Build GRCh38
rs1042713822 Chr.21: 30289364-30289364 on Build GRCh38
rs754400383 Chr.21: 30289278-30289278 on Build GRCh38

View Full Product Details