Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 83462624-83462895 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM175A
Entrez Gene ID
84142
Gene Name
family with sequence similarity 175 member A
Gene Aliases
ABRA1, CCDC98
UniGene
Hs.334772
Gene Chromosome Location
Chr.4: 83459515-83485181 on Build GRCh38
Transcripts
NM_139076.2 XM_005263280.4 XM_017008694.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6919224 c.904A>C p.K302Q Chr.4: 83462795-83462795 on Build GRCh38
6210940 c.1000G>A p.E334K Chr.4: 83462699-83462699 on Build GRCh38
7296716 c.804G>A p.K268K Chr.4: 83462895-83462895 on Build GRCh38
6956398 c.908delA p.N303fs*17 Chr.4: 83462791-83462791 on Build GRCh38
3760790 c.1042G>A p.A348T Chr.4: 83462657-83462657 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760360071 Chr.4: 83462779-83462779 on Build GRCh38
rs758777622 Chr.4: 83462783-83462783 on Build GRCh38
rs754103619 Chr.4: 83462830-83462830 on Build GRCh38
rs749943018 Chr.4: 83462774-83462774 on Build GRCh38
rs35378950 Chr.4: 83462835-83462835 on Build GRCh38
rs752068094 Chr.4: 83462795-83462795 on Build GRCh38
rs778357689 Chr.4: 83462713-83462713 on Build GRCh38
rs960772904 Chr.4: 83462867-83462867 on Build GRCh38
rs587780267 Chr.4: 83462866-83462866 on Build GRCh38
rs900639769 Chr.4: 83462694-83462694 on Build GRCh38
rs781307588 Chr.4: 83462803-83462803 on Build GRCh38
rs779312880 Chr.4: 83462669-83462669 on Build GRCh38
rs771082457 Chr.4: 83462734-83462734 on Build GRCh38
rs775345294 Chr.4: 83462747-83462747 on Build GRCh38
rs774216794 Chr.4: 83462764-83462764 on Build GRCh38
rs747527820 Chr.4: 83462718-83462718 on Build GRCh38
rs780076912 Chr.4: 83462845-83462845 on Build GRCh38
rs764429803 Chr.4: 83462688-83462688 on Build GRCh38
rs766975460 Chr.4: 83462771-83462771 on Build GRCh38
rs748047951 Chr.4: 83462864-83462864 on Build GRCh38
rs377162113 Chr.4: 83462809-83462809 on Build GRCh38
rs781160014 Chr.4: 83462740-83462740 on Build GRCh38
rs748475674 Chr.4: 83462871-83462873 on Build GRCh38
rs140407442 Chr.4: 83462802-83462802 on Build GRCh38
rs376489465 Chr.4: 83462758-83462758 on Build GRCh38
rs181090081 Chr.4: 83462868-83462868 on Build GRCh38
rs745996212 Chr.4: 83462810-83462810 on Build GRCh38
rs587780268 Chr.4: 83462863-83462863 on Build GRCh38
rs12642536 Chr.4: 83462657-83462657 on Build GRCh38
rs143012786 Chr.4: 83462804-83462804 on Build GRCh38
rs367958380 Chr.4: 83462680-83462680 on Build GRCh38
rs79357787 Chr.4: 83462748-83462748 on Build GRCh38
rs746068286 Chr.4: 83462742-83462742 on Build GRCh38
rs758969039 Chr.4: 83462681-83462681 on Build GRCh38
rs755755614 Chr.4: 83462663-83462663 on Build GRCh38
rs778821977 Chr.4: 83462858-83462861 on Build GRCh38
rs769078691 Chr.4: 83462819-83462820 on Build GRCh38
rs757188204 Chr.4: 83462834-83462835 on Build GRCh38
rs778300694 Chr.4: 83462787-83462787 on Build GRCh38
rs766011574 Chr.4: 83462781-83462781 on Build GRCh38
rs150371881 Chr.4: 83462846-83462846 on Build GRCh38
rs587780261 Chr.4: 83462666-83462667 on Build GRCh38
rs751068863 Chr.4: 83462651-83462651 on Build GRCh38
rs138986552 Chr.4: 83462782-83462782 on Build GRCh38
rs756357242 Chr.4: 83462831-83462831 on Build GRCh38
rs761761558 Chr.4: 83462767-83462767 on Build GRCh38
rs750046185 Chr.4: 83462661-83462661 on Build GRCh38
rs587780269 Chr.4: 83462784-83462784 on Build GRCh38
rs769962886 Chr.4: 83462746-83462746 on Build GRCh38
rs768392156 Chr.4: 83462850-83462850 on Build GRCh38

View Full Product Details