Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 159037987-159038248 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAGAP
Entrez Gene ID
117289
Gene Name
T-cell activation RhoGTPase activating protein
Gene Aliases
ARHGAP47, FKSG15, IDDM21, TAGAP1
UniGene
Hs.529984
Gene Chromosome Location
Chr.6: 159034468-159045152 on Build GRCh38
Transcripts
NM_001278733.1 NM_054114.4 NM_152133.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6413301 c.870T>A p.D290E Chr.6: 159038142-159038142 on Build GRCh38
3728494 c.833G>T p.G278V Chr.6: 159038179-159038179 on Build GRCh38
6392998 c.896C>T p.S299L Chr.6: 159038116-159038116 on Build GRCh38
1267384 c.811G>C p.D271H Chr.6: 159038201-159038201 on Build GRCh38
1228456 c.814A>G p.N272D Chr.6: 159038198-159038198 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756136147 Chr.6: 159038199-159038199 on Build GRCh38
rs754842183 Chr.6: 159038210-159038210 on Build GRCh38
rs753717176 Chr.6: 159038208-159038208 on Build GRCh38
rs373695805 Chr.6: 159038196-159038196 on Build GRCh38
rs560226623 Chr.6: 159038216-159038216 on Build GRCh38
rs774098883 Chr.6: 159038149-159038149 on Build GRCh38
rs747518640 Chr.6: 159038121-159038121 on Build GRCh38
rs201081350 Chr.6: 159038072-159038072 on Build GRCh38
rs568144827 Chr.6: 159038057-159038057 on Build GRCh38
rs376139692 Chr.6: 159038102-159038102 on Build GRCh38
rs115774479 Chr.6: 159038018-159038018 on Build GRCh38
rs556418748 Chr.6: 159038065-159038065 on Build GRCh38
rs539913868 Chr.6: 159038203-159038203 on Build GRCh38
rs370261958 Chr.6: 159038115-159038115 on Build GRCh38
rs577701787 Chr.6: 159038122-159038122 on Build GRCh38
rs761505429 Chr.6: 159038161-159038161 on Build GRCh38
rs372975422 Chr.6: 159038079-159038079 on Build GRCh38
rs765484870 Chr.6: 159038145-159038145 on Build GRCh38
rs776988940 Chr.6: 159038124-159038124 on Build GRCh38
rs760010198 Chr.6: 159038126-159038126 on Build GRCh38
rs112913154 Chr.6: 159038162-159038162 on Build GRCh38
rs934379119 Chr.6: 159038167-159038167 on Build GRCh38
rs778550964 Chr.6: 159038214-159038214 on Build GRCh38
rs758549217 Chr.6: 159038219-159038219 on Build GRCh38
rs948731632 Chr.6: 159038180-159038180 on Build GRCh38
rs748287566 Chr.6: 159038087-159038087 on Build GRCh38
rs930609080 Chr.6: 159038050-159038050 on Build GRCh38
rs1016064537 Chr.6: 159038076-159038076 on Build GRCh38
rs539681147 Chr.6: 159038041-159038041 on Build GRCh38

View Full Product Details