Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6516791-6517064 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCAPD2
Entrez Gene ID
9918
Gene Name
non-SMC condensin I complex subunit D2
Gene Aliases
CAP-D2, CNAP1, hCAP-D2
UniGene
Hs.5719
Gene Chromosome Location
Chr.12: 6494132-6531966 on Build GRCh38
Transcripts
NM_014865.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
223514 c.1066G>A p.A356T Chr.12: 6516906-6516906 on Build GRCh38
6855680 c.1025C>T p.A342V Chr.12: 6516865-6516865 on Build GRCh38
6722700 c.1088A>G p.D363G Chr.12: 6516928-6516928 on Build GRCh38
6519920 c.1116T>A p.H372Q Chr.12: 6516956-6516956 on Build GRCh38
4643756 c.1132T>G p.S378A Chr.12: 6516972-6516972 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202243696 Chr.12: 6516980-6516980 on Build GRCh38
rs537119663 Chr.12: 6516861-6516861 on Build GRCh38
rs762076491 Chr.12: 6516922-6516922 on Build GRCh38
rs747533042 Chr.12: 6517028-6517028 on Build GRCh38
rs769823890 Chr.12: 6517039-6517039 on Build GRCh38
rs141122541 Chr.12: 6517033-6517033 on Build GRCh38
rs750204852 Chr.12: 6516829-6516829 on Build GRCh38
rs780431663 Chr.12: 6517021-6517021 on Build GRCh38
rs762678605 Chr.12: 6516840-6516840 on Build GRCh38
rs566110893 Chr.12: 6516956-6516956 on Build GRCh38
rs771302335 Chr.12: 6516841-6516841 on Build GRCh38
rs778053909 Chr.12: 6516972-6516972 on Build GRCh38
rs369341180 Chr.12: 6517009-6517009 on Build GRCh38
rs773356519 Chr.12: 6516940-6516940 on Build GRCh38
rs772501599 Chr.12: 6516883-6516883 on Build GRCh38
rs779788249 Chr.12: 6516882-6516883 on Build GRCh38
rs147023142 Chr.12: 6516872-6516872 on Build GRCh38
rs752238777 Chr.12: 6516983-6516983 on Build GRCh38
rs886280771 Chr.12: 6516938-6516938 on Build GRCh38
rs763847471 Chr.12: 6517007-6517007 on Build GRCh38
rs755825630 Chr.12: 6516875-6516875 on Build GRCh38
rs377724145 Chr.12: 6516816-6516817 on Build GRCh38
rs781501861 Chr.12: 6516865-6516865 on Build GRCh38
rs762060397 Chr.12: 6516823-6516823 on Build GRCh38
rs748231558 Chr.12: 6517034-6517034 on Build GRCh38
rs776764263 Chr.12: 6516916-6516916 on Build GRCh38
rs767003306 Chr.12: 6516982-6516982 on Build GRCh38
rs770015862 Chr.12: 6516939-6516939 on Build GRCh38
rs1027786615 Chr.12: 6516900-6516900 on Build GRCh38
rs779481219 Chr.12: 6516877-6516877 on Build GRCh38
rs748279493 Chr.12: 6516888-6516888 on Build GRCh38
rs780733818 Chr.12: 6516866-6516866 on Build GRCh38
rs746799615 Chr.12: 6516941-6516945 on Build GRCh38
rs758153209 Chr.12: 6516825-6516825 on Build GRCh38
rs768454744 Chr.12: 6516988-6516989 on Build GRCh38
rs759487830 Chr.12: 6516981-6516981 on Build GRCh38
rs756476624 Chr.12: 6516963-6516963 on Build GRCh38
rs1016312186 Chr.12: 6516925-6516925 on Build GRCh38
rs143795587 Chr.12: 6516849-6516849 on Build GRCh38
rs138198563 Chr.12: 6516913-6516913 on Build GRCh38
rs755493259 Chr.12: 6517029-6517029 on Build GRCh38
rs994613441 Chr.12: 6516894-6516894 on Build GRCh38
rs768655520 Chr.12: 6516915-6516915 on Build GRCh38
rs576859525 Chr.12: 6517006-6517006 on Build GRCh38
rs370387241 Chr.12: 6516895-6516895 on Build GRCh38
rs762766576 Chr.12: 6516941-6516941 on Build GRCh38
rs201334852 Chr.12: 6516899-6516899 on Build GRCh38

View Full Product Details