Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 168924824-168925065 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBR4
Entrez Gene ID
84869
Gene Name
carbonyl reductase 4
Gene Aliases
SDR45C1
UniGene
Hs.659311
Gene Chromosome Location
Chr.4: 168894484-169010317 on Build GRCh38
Transcripts
XM_005263315.2 XM_017008782.1 NM_001166108.1 NM_001166109.1 NM_001166110.1 NM_016081.3 XM_005262861.4 XM_005262866.2 XM_011531768.2 XM_011531769.2 XM_011531770.2 XM_011531771.2 XM_011531772.2 XM_011531773.1 XM_011531774.1 XM_011531775.1 XM_017007910.1

Gene Symbol
PALLD
Entrez Gene ID
23022
Gene Name
palladin, cytoskeletal associated protein
Gene Aliases
CGI-151, CGI151, MYN, PNCA1, SIH002
UniGene
Hs.151220
Gene Chromosome Location
Chr.4: 168497039-168928457 on Build GRCh38
Transcripts
XM_005263315.2 XM_017008782.1 NM_001166108.1 NM_001166109.1 NM_001166110.1 NM_016081.3 XM_005262861.4 XM_005262866.2 XM_011531768.2 XM_011531769.2 XM_011531770.2 XM_011531771.2 XM_011531772.2 XM_011531773.1 XM_011531774.1 XM_011531775.1 XM_017007910.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3601983 c.3216G>T p.Q1072H Chr.4: 168924987-168924987 on Build GRCh38
4978527 c.3203G>A p.C1068Y Chr.4: 168924974-168924974 on Build GRCh38
3601986 c.2743G>A p.G915R Chr.4: 168924988-168924988 on Build GRCh38
257663 c.3290C>T p.A1097V Chr.4: 168925061-168925061 on Build GRCh38
6307308 c.3218G>T p.G1073V Chr.4: 168924989-168924989 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs587780756 Chr.4: 168924989-168924989 on Build GRCh38
rs771679229 Chr.4: 168924974-168924974 on Build GRCh38
rs745362518 Chr.4: 168924964-168924964 on Build GRCh38
rs779615947 Chr.4: 168924963-168924963 on Build GRCh38
rs781692525 Chr.4: 168924910-168924911 on Build GRCh38
rs753025711 Chr.4: 168924918-168924918 on Build GRCh38
rs114593924 Chr.4: 168925017-168925017 on Build GRCh38
rs752802030 Chr.4: 168925029-168925029 on Build GRCh38
rs35175376 Chr.4: 168924965-168924966 on Build GRCh38
rs996863687 Chr.4: 168924877-168924877 on Build GRCh38
rs764265611 Chr.4: 168925041-168925041 on Build GRCh38
rs983488446 Chr.4: 168924855-168924855 on Build GRCh38
rs765135925 Chr.4: 168925003-168925003 on Build GRCh38
rs767636400 Chr.4: 168925023-168925023 on Build GRCh38
rs761634920 Chr.4: 168924999-168924999 on Build GRCh38
rs907990778 Chr.4: 168924905-168924905 on Build GRCh38
rs776518808 Chr.4: 168924996-168924996 on Build GRCh38
rs749775897 Chr.4: 168924957-168924957 on Build GRCh38
rs1027467560 Chr.4: 168924909-168924909 on Build GRCh38
rs150256686 Chr.4: 168924978-168924978 on Build GRCh38
rs748489494 Chr.4: 168925031-168925032 on Build GRCh38
rs771536528 Chr.4: 168924958-168924958 on Build GRCh38
rs587780201 Chr.4: 168924984-168924984 on Build GRCh38
rs939449621 Chr.4: 168924922-168924922 on Build GRCh38
rs59633770 Chr.4: 168924976-168924976 on Build GRCh38
rs756392060 Chr.4: 168925040-168925040 on Build GRCh38
rs778210310 Chr.4: 168924949-168924949 on Build GRCh38
rs757105205 Chr.4: 168924885-168924885 on Build GRCh38
rs368806050 Chr.4: 168925021-168925021 on Build GRCh38
rs756363802 Chr.4: 168924924-168924924 on Build GRCh38

View Full Product Details