Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 17022529-17022802 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SDHB
Entrez Gene ID
6390
Gene Name
succinate dehydrogenase complex iron sulfur subunit B
Gene Aliases
CWS2, IP, PGL4, SDH, SDH1, SDH2, SDHIP
UniGene
Hs.465924
Gene Chromosome Location
Chr.1: 17018722-17054170 on Build GRCh38
Transcripts
NM_003000.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6982382 c.752G>A p.R251K Chr.1: 17022621-17022621 on Build GRCh38
5613817 c.710C>T p.P237L Chr.1: 17022663-17022663 on Build GRCh38
899738 c.688C>T p.R230C Chr.1: 17022685-17022685 on Build GRCh38
6914218 c.734C>T p.T245I Chr.1: 17022639-17022639 on Build GRCh38
4964107 c.719T>A p.L240Q Chr.1: 17022654-17022654 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747518441 Chr.1: 17022723-17022723 on Build GRCh38
rs878854578 Chr.1: 17022714-17022714 on Build GRCh38
rs940695306 Chr.1: 17022775-17022775 on Build GRCh38
rs376106876 Chr.1: 17022766-17022766 on Build GRCh38
rs748796588 Chr.1: 17022734-17022734 on Build GRCh38
rs587782604 Chr.1: 17022684-17022684 on Build GRCh38
rs370397369 Chr.1: 17022671-17022671 on Build GRCh38
rs201728852 Chr.1: 17022673-17022673 on Build GRCh38
rs779143585 Chr.1: 17022677-17022677 on Build GRCh38
rs201843552 Chr.1: 17022779-17022779 on Build GRCh38
rs889343638 Chr.1: 17022574-17022574 on Build GRCh38
rs781329558 Chr.1: 17022564-17022564 on Build GRCh38
rs767433861 Chr.1: 17022778-17022778 on Build GRCh38
rs749542793 Chr.1: 17022580-17022580 on Build GRCh38
rs115561881 Chr.1: 17022595-17022595 on Build GRCh38
rs771136851 Chr.1: 17022586-17022586 on Build GRCh38
rs786203251 Chr.1: 17022649-17022649 on Build GRCh38
rs747898945 Chr.1: 17022556-17022556 on Build GRCh38
rs752795496 Chr.1: 17022565-17022565 on Build GRCh38
rs200245469 Chr.1: 17022724-17022724 on Build GRCh38
rs769359897 Chr.1: 17022618-17022618 on Build GRCh38
rs786202913 Chr.1: 17022672-17022672 on Build GRCh38
rs41273161 Chr.1: 17022579-17022579 on Build GRCh38
rs748333885 Chr.1: 17022783-17022783 on Build GRCh38
rs878854583 Chr.1: 17022650-17022650 on Build GRCh38
rs146800605 Chr.1: 17022637-17022637 on Build GRCh38
rs876660642 Chr.1: 17022660-17022660 on Build GRCh38
rs374921867 Chr.1: 17022737-17022737 on Build GRCh38
rs201832634 Chr.1: 17022599-17022599 on Build GRCh38
rs74315368 Chr.1: 17022648-17022648 on Build GRCh38
rs186768244 Chr.1: 17022664-17022664 on Build GRCh38
rs762812025 Chr.1: 17022689-17022690 on Build GRCh38
rs199925530 Chr.1: 17022780-17022780 on Build GRCh38
rs756329877 Chr.1: 17022568-17022568 on Build GRCh38
rs794728950 Chr.1: 17022652-17022655 on Build GRCh38
rs774215357 Chr.1: 17022746-17022746 on Build GRCh38
rs916516745 Chr.1: 17022719-17022719 on Build GRCh38
rs587781266 Chr.1: 17022654-17022657 on Build GRCh38
rs201923876 Chr.1: 17022563-17022563 on Build GRCh38
rs201094649 Chr.1: 17022620-17022620 on Build GRCh38
rs780477085 Chr.1: 17022703-17022703 on Build GRCh38
rs786201085 Chr.1: 17022615-17022615 on Build GRCh38
rs201019634 Chr.1: 17022776-17022776 on Build GRCh38
rs772798766 Chr.1: 17022730-17022730 on Build GRCh38
rs34309090 Chr.1: 17022612-17022612 on Build GRCh38
rs764034093 Chr.1: 17022781-17022781 on Build GRCh38
rs199945904 Chr.1: 17022603-17022603 on Build GRCh38
rs909245917 Chr.1: 17022764-17022764 on Build GRCh38
rs201098090 Chr.1: 17022657-17022657 on Build GRCh38
rs948484408 Chr.1: 17022612-17022612 on Build GRCh38
rs1057517537 Chr.1: 17022624-17022624 on Build GRCh38
rs776039903 Chr.1: 17022604-17022604 on Build GRCh38
rs766038910 Chr.1: 17022643-17022643 on Build GRCh38
rs200896502 Chr.1: 17022634-17022634 on Build GRCh38
rs778914018 Chr.1: 17022554-17022554 on Build GRCh38
rs759446340 Chr.1: 17022653-17022653 on Build GRCh38
rs969680248 Chr.1: 17022755-17022755 on Build GRCh38
rs984814872 Chr.1: 17022758-17022758 on Build GRCh38
rs746277655 Chr.1: 17022596-17022596 on Build GRCh38
rs761295633 Chr.1: 17022608-17022608 on Build GRCh38
rs878854582 Chr.1: 17022651-17022651 on Build GRCh38
rs752574758 Chr.1: 17022567-17022592 on Build GRCh38
rs138996609 Chr.1: 17022685-17022685 on Build GRCh38
rs878854579 Chr.1: 17022663-17022663 on Build GRCh38
rs746224555 Chr.1: 17022678-17022678 on Build GRCh38
rs897839811 Chr.1: 17022752-17022752 on Build GRCh38

View Full Product Details