Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 12351504-12351759 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPARG
Entrez Gene ID
5468
Gene Name
peroxisome proliferator activated receptor gamma
Gene Aliases
CIMT1, GLM1, NR1C3, PPARG1, PPARG2, PPARgamma
UniGene
Hs.162646
Gene Chromosome Location
Chr.3: 12287850-12471054 on Build GRCh38
Transcripts
NM_005037.5 NM_015869.4 NM_138711.3 NM_138712.3 XM_011533841.2 XM_011533842.2 XM_011533843.2 XM_011533844.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4987222 c.34C>G p.P12A Chr.3: 12351626-12351626 on Build GRCh38
7102682 c.57_58insA p.T20fs*14 Chr.3: 12351649-12351650 on Build GRCh38
581315 c.7G>A p.E3K Chr.3: 12351599-12351599 on Build GRCh38
26993 c.11C>G p.T4S Chr.3: 12351603-12351603 on Build GRCh38
1692394 c.77C>T p.S26L Chr.3: 12351669-12351669 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111473633 Chr.3: 12351713-12351713 on Build GRCh38
rs757329632 Chr.3: 12351543-12351543 on Build GRCh38
rs370961244 Chr.3: 12351575-12351575 on Build GRCh38
rs565261168 Chr.3: 12351701-12351701 on Build GRCh38
rs943725719 Chr.3: 12351635-12351635 on Build GRCh38
rs765473307 Chr.3: 12351623-12351623 on Build GRCh38
rs765281286 Chr.3: 12351547-12351547 on Build GRCh38
rs373988251 Chr.3: 12351576-12351576 on Build GRCh38
rs775628426 Chr.3: 12351593-12351593 on Build GRCh38
rs193239395 Chr.3: 12351708-12351708 on Build GRCh38
rs184470181 Chr.3: 12351714-12351714 on Build GRCh38
rs928058229 Chr.3: 12351605-12351605 on Build GRCh38
rs745378751 Chr.3: 12351587-12351587 on Build GRCh38
rs199970439 Chr.3: 12351695-12351695 on Build GRCh38
rs375584938 Chr.3: 12351699-12351699 on Build GRCh38
rs752068471 Chr.3: 12351643-12351643 on Build GRCh38
rs200479885 Chr.3: 12351571-12351571 on Build GRCh38
rs759923616 Chr.3: 12351722-12351722 on Build GRCh38
rs776891749 Chr.3: 12351620-12351620 on Build GRCh38
rs1055170892 Chr.3: 12351633-12351633 on Build GRCh38
rs768377660 Chr.3: 12351614-12351614 on Build GRCh38
rs1801282 Chr.3: 12351626-12351626 on Build GRCh38
rs572991054 Chr.3: 12351653-12351653 on Build GRCh38
rs112653857 Chr.3: 12351694-12351694 on Build GRCh38
rs750523320 Chr.3: 12351548-12351548 on Build GRCh38
rs367903978 Chr.3: 12351690-12351690 on Build GRCh38
rs1040659276 Chr.3: 12351640-12351640 on Build GRCh38
rs763032994 Chr.3: 12351632-12351632 on Build GRCh38
rs773477350 Chr.3: 12351707-12351707 on Build GRCh38
rs772080186 Chr.3: 12351588-12351588 on Build GRCh38
rs961425304 Chr.3: 12351537-12351537 on Build GRCh38
rs1026032168 Chr.3: 12351703-12351703 on Build GRCh38
rs748929397 Chr.3: 12351580-12351580 on Build GRCh38
rs758861013 Chr.3: 12351550-12351550 on Build GRCh38
rs145161478 Chr.3: 12351634-12351634 on Build GRCh38
rs771775640 Chr.3: 12351693-12351693 on Build GRCh38
rs753139831 Chr.3: 12351668-12351668 on Build GRCh38
rs778529753 Chr.3: 12351584-12351584 on Build GRCh38
rs35206526 Chr.3: 12351542-12351545 on Build GRCh38
rs761891193 Chr.3: 12351622-12351622 on Build GRCh38
rs368662399 Chr.3: 12351700-12351700 on Build GRCh38
rs752081192 Chr.3: 12351715-12351715 on Build GRCh38
rs575440954 Chr.3: 12351564-12351564 on Build GRCh38
rs760515031 Chr.3: 12351607-12351607 on Build GRCh38
rs745362327 Chr.3: 12351691-12351691 on Build GRCh38
rs756794860 Chr.3: 12351678-12351678 on Build GRCh38
rs755335866 Chr.3: 12351649-12351649 on Build GRCh38
rs560683642 Chr.3: 12351639-12351639 on Build GRCh38
rs534301709 Chr.3: 12351538-12351541 on Build GRCh38
rs772801073 Chr.3: 12351690-12351690 on Build GRCh38

View Full Product Details