Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 186296641-186296914 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRG4
Entrez Gene ID
10216
Gene Name
proteoglycan 4
Gene Aliases
CACP, HAPO, JCAP, MSF, SZP
UniGene
Hs.647723
Gene Chromosome Location
Chr.1: 186296273-186314562 on Build GRCh38
Transcripts
NM_001127708.2 NM_001127709.2 NM_001127710.2 NM_001303232.1 NM_005807.4 XM_017000002.1 XM_017000003.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6486688 c.2T>C p.M1T Chr.1: 186296877-186296877 on Build GRCh38
1473165 c.26A>C p.Y9S Chr.1: 186296901-186296901 on Build GRCh38
1320639 c.30G>T p.L10L Chr.1: 186296905-186296905 on Build GRCh38
3723191 c.19C>A p.P7T Chr.1: 186296894-186296894 on Build GRCh38
3863871 c.21C>T p.P7P Chr.1: 186296896-186296896 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs892523778 Chr.1: 186296722-186296722 on Build GRCh38
rs78126824 Chr.1: 186296749-186296749 on Build GRCh38
rs568736879 Chr.1: 186296681-186296681 on Build GRCh38
rs1002645844 Chr.1: 186296668-186296668 on Build GRCh38
rs774599749 Chr.1: 186296859-186296859 on Build GRCh38
rs903684595 Chr.1: 186296679-186296679 on Build GRCh38
rs143556920 Chr.1: 186296879-186296879 on Build GRCh38
rs1049829922 Chr.1: 186296863-186296863 on Build GRCh38
rs139171967 Chr.1: 186296874-186296874 on Build GRCh38
rs149511072 Chr.1: 186296875-186296875 on Build GRCh38
rs780651425 Chr.1: 186296844-186296844 on Build GRCh38
rs550079037 Chr.1: 186296673-186296673 on Build GRCh38
rs200387997 Chr.1: 186296858-186296858 on Build GRCh38
rs770412448 Chr.1: 186296843-186296843 on Build GRCh38
rs762263779 Chr.1: 186296774-186296774 on Build GRCh38
rs529791040 Chr.1: 186296887-186296887 on Build GRCh38
rs553592821 Chr.1: 186296697-186296697 on Build GRCh38
rs769070129 Chr.1: 186296829-186296833 on Build GRCh38
rs752562170 Chr.1: 186296876-186296876 on Build GRCh38
rs10911844 Chr.1: 186296824-186296824 on Build GRCh38
rs908684029 Chr.1: 186296735-186296735 on Build GRCh38
rs777159659 Chr.1: 186296829-186296829 on Build GRCh38
rs79536913 Chr.1: 186296808-186296808 on Build GRCh38
rs190597799 Chr.1: 186296882-186296882 on Build GRCh38
rs576427419 Chr.1: 186296891-186296891 on Build GRCh38
rs1293987 Chr.1: 186296816-186296816 on Build GRCh38
rs34444883 Chr.1: 186296679-186296679 on Build GRCh38
rs889405557 Chr.1: 186296783-186296783 on Build GRCh38
rs1052967739 Chr.1: 186296721-186296721 on Build GRCh38
rs992375453 Chr.1: 186296734-186296734 on Build GRCh38
rs749820846 Chr.1: 186296851-186296851 on Build GRCh38
rs751122318 Chr.1: 186296881-186296881 on Build GRCh38
rs569955781 Chr.1: 186296749-186296750 on Build GRCh38
rs1016463951 Chr.1: 186296812-186296812 on Build GRCh38
rs1033698526 Chr.1: 186296683-186296683 on Build GRCh38
rs772375510 Chr.1: 186296864-186296864 on Build GRCh38

View Full Product Details