Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 20875344-20875579 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLCO1B3
Entrez Gene ID
28234
Gene Name
solute carrier organic anion transporter family member 1B3
Gene Aliases
HBLRR, LST-2, LST-3TM13, LST3, OATP-8, OATP1B3, OATP8, SLC21A8
UniGene
Hs.504966
Gene Chromosome Location
Chr.12: 20810704-20916911 on Build GRCh38
Transcripts
NM_019844.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3459116 c.884C>T p.S295L Chr.12: 20875391-20875391 on Build GRCh38
692856 c.840G>A p.P280P Chr.12: 20875347-20875347 on Build GRCh38
4438445 c.837G>T p.L279F Chr.12: 20875344-20875344 on Build GRCh38
3459115 c.865G>A p.E289K Chr.12: 20875372-20875372 on Build GRCh38
1705283 c.878C>T p.S293L Chr.12: 20875385-20875385 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774984457 Chr.12: 20875450-20875450 on Build GRCh38
rs768387734 Chr.12: 20875454-20875454 on Build GRCh38
rs773440798 Chr.12: 20875463-20875463 on Build GRCh38
rs762437057 Chr.12: 20875478-20875478 on Build GRCh38
rs1037936412 Chr.12: 20875541-20875541 on Build GRCh38
rs760723958 Chr.12: 20875469-20875469 on Build GRCh38
rs756852295 Chr.12: 20875408-20875408 on Build GRCh38
rs754356216 Chr.12: 20875397-20875397 on Build GRCh38
rs779597759 Chr.12: 20875529-20875529 on Build GRCh38
rs916670665 Chr.12: 20875466-20875466 on Build GRCh38
rs946671511 Chr.12: 20875481-20875481 on Build GRCh38
rs200068079 Chr.12: 20875379-20875379 on Build GRCh38
rs754342222 Chr.12: 20875473-20875473 on Build GRCh38
rs756903628 Chr.12: 20875496-20875496 on Build GRCh38
rs543399361 Chr.12: 20875519-20875519 on Build GRCh38
rs1033345418 Chr.12: 20875386-20875386 on Build GRCh38
rs769780056 Chr.12: 20875420-20875420 on Build GRCh38
rs983726989 Chr.12: 20875419-20875419 on Build GRCh38
rs777927970 Chr.12: 20875465-20875465 on Build GRCh38
rs765610427 Chr.12: 20875482-20875482 on Build GRCh38
rs142873062 Chr.12: 20875380-20875380 on Build GRCh38
rs979472298 Chr.12: 20875487-20875487 on Build GRCh38
rs755946165 Chr.12: 20875451-20875452 on Build GRCh38
rs757534726 Chr.12: 20875486-20875495 on Build GRCh38
rs117522823 Chr.12: 20875498-20875498 on Build GRCh38
rs772355903 Chr.12: 20875457-20875457 on Build GRCh38
rs755365122 Chr.12: 20875399-20875399 on Build GRCh38
rs373072758 Chr.12: 20875455-20875455 on Build GRCh38
rs780708027 Chr.12: 20875418-20875418 on Build GRCh38
rs778994317 Chr.12: 20875515-20875520 on Build GRCh38
rs752635362 Chr.12: 20875429-20875430 on Build GRCh38
rs1027879448 Chr.12: 20875417-20875417 on Build GRCh38
rs766475606 Chr.12: 20875470-20875470 on Build GRCh38
rs371298832 Chr.12: 20875442-20875442 on Build GRCh38
rs753017803 Chr.12: 20875495-20875495 on Build GRCh38
rs142909652 Chr.12: 20875536-20875537 on Build GRCh38
rs751260776 Chr.12: 20875403-20875403 on Build GRCh38
rs933769093 Chr.12: 20875432-20875432 on Build GRCh38
rs370474386 Chr.12: 20875528-20875528 on Build GRCh38
rs753817906 Chr.12: 20875464-20875465 on Build GRCh38
rs779304579 Chr.12: 20875402-20875402 on Build GRCh38
rs766738737 Chr.12: 20875395-20875395 on Build GRCh38
rs750076005 Chr.12: 20875523-20875523 on Build GRCh38
rs994516947 Chr.12: 20875389-20875389 on Build GRCh38
rs988505234 Chr.12: 20875392-20875392 on Build GRCh38
rs146565174 Chr.12: 20875456-20875456 on Build GRCh38

View Full Product Details