Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 158700216-158700485 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR6K2
Entrez Gene ID
81448
Gene Name
olfactory receptor family 6 subfamily K member 2
Gene Aliases
OR1-17
UniGene
Hs.554489
Gene Chromosome Location
Chr.1: 158699678-158700652 on Build GRCh38
Transcripts
NM_001005279.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
293202 c.318C>T p.T106T Chr.1: 158700335-158700335 on Build GRCh38
414402 c.207G>C p.L69L Chr.1: 158700446-158700446 on Build GRCh38
6478215 c.249T>A p.S83S Chr.1: 158700404-158700404 on Build GRCh38
4023824 c.423C>A p.T141T Chr.1: 158700230-158700230 on Build GRCh38
6459468 c.363C>A p.H121Q Chr.1: 158700290-158700290 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139620465 Chr.1: 158700274-158700274 on Build GRCh38
rs558029562 Chr.1: 158700307-158700307 on Build GRCh38
rs781434431 Chr.1: 158700355-158700355 on Build GRCh38
rs76691067 Chr.1: 158700257-158700257 on Build GRCh38
rs370003896 Chr.1: 158700288-158700288 on Build GRCh38
rs777624103 Chr.1: 158700314-158700314 on Build GRCh38
rs768885609 Chr.1: 158700343-158700343 on Build GRCh38
rs895567075 Chr.1: 158700359-158700359 on Build GRCh38
rs377720257 Chr.1: 158700311-158700311 on Build GRCh38
rs142441693 Chr.1: 158700362-158700362 on Build GRCh38
rs762384880 Chr.1: 158700315-158700315 on Build GRCh38
rs994286088 Chr.1: 158700397-158700397 on Build GRCh38
rs766846896 Chr.1: 158700303-158700303 on Build GRCh38
rs369302511 Chr.1: 158700435-158700435 on Build GRCh38
rs561477081 Chr.1: 158700432-158700432 on Build GRCh38
rs754692467 Chr.1: 158700306-158700306 on Build GRCh38
rs146014021 Chr.1: 158700405-158700405 on Build GRCh38
rs774876083 Chr.1: 158700392-158700392 on Build GRCh38
rs145450386 Chr.1: 158700453-158700453 on Build GRCh38
rs371952569 Chr.1: 158700285-158700286 on Build GRCh38
rs773659942 Chr.1: 158700370-158700370 on Build GRCh38
rs760156630 Chr.1: 158700422-158700422 on Build GRCh38
rs776703481 Chr.1: 158700286-158700286 on Build GRCh38
rs764256515 Chr.1: 158700272-158700273 on Build GRCh38
rs751906532 Chr.1: 158700304-158700304 on Build GRCh38
rs77517860 Chr.1: 158700430-158700430 on Build GRCh38
rs1045256493 Chr.1: 158700394-158700394 on Build GRCh38
rs759937297 Chr.1: 158700401-158700401 on Build GRCh38
rs770576605 Chr.1: 158700335-158700335 on Build GRCh38
rs374289998 Chr.1: 158700294-158700294 on Build GRCh38
rs568479215 Chr.1: 158700328-158700328 on Build GRCh38
rs150717680 Chr.1: 158700428-158700429 on Build GRCh38
rs776690962 Chr.1: 158700352-158700352 on Build GRCh38
rs753805288 Chr.1: 158700316-158700316 on Build GRCh38
rs752182709 Chr.1: 158700309-158700309 on Build GRCh38
rs750251784 Chr.1: 158700433-158700433 on Build GRCh38
rs773736572 Chr.1: 158700302-158700302 on Build GRCh38
rs781319145 Chr.1: 158700445-158700445 on Build GRCh38
rs753223476 Chr.1: 158700429-158700429 on Build GRCh38
rs868375262 Chr.1: 158700356-158700356 on Build GRCh38
rs756030265 Chr.1: 158700246-158700246 on Build GRCh38
rs977143592 Chr.1: 158700243-158700243 on Build GRCh38
rs771505818 Chr.1: 158700391-158700391 on Build GRCh38
rs748218001 Chr.1: 158700361-158700361 on Build GRCh38
rs770694846 Chr.1: 158700270-158700270 on Build GRCh38
rs746826865 Chr.1: 158700285-158700285 on Build GRCh38
rs778681132 Chr.1: 158700334-158700334 on Build GRCh38
rs572481654 Chr.1: 158700276-158700276 on Build GRCh38
rs757107613 Chr.1: 158700326-158700326 on Build GRCh38
rs752369306 Chr.1: 158700416-158700416 on Build GRCh38
rs888591448 Chr.1: 158700408-158700408 on Build GRCh38
rs913038867 Chr.1: 158700300-158700300 on Build GRCh38
rs769896487 Chr.1: 158700363-158700363 on Build GRCh38
rs763659944 Chr.1: 158700424-158700424 on Build GRCh38
rs763557435 Chr.1: 158700390-158700390 on Build GRCh38
rs201819817 Chr.1: 158700250-158700250 on Build GRCh38

View Full Product Details