Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 36929549-36929822 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSF2RB
Entrez Gene ID
1439
Gene Name
colony stimulating factor 2 receptor beta common subunit
Gene Aliases
CD131, CDw131, IL3RB, IL5RB, SMDP5
UniGene
Hs.592192
Gene Chromosome Location
Chr.22: 36913574-36940449 on Build GRCh38
Transcripts
NM_000395.2 XM_005261340.3 XM_011529903.2 XM_011529904.2 XM_011529905.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
444946 c.631C>T p.R211* Chr.22: 36929720-36929720 on Build GRCh38
1033899 c.628G>A p.A210T Chr.22: 36929717-36929717 on Build GRCh38
3554021 c.629C>T p.A210V Chr.22: 36929718-36929718 on Build GRCh38
6636120 c.549G>A p.E183E Chr.22: 36929559-36929559 on Build GRCh38
5810242 c.679A>T p.S227C Chr.22: 36929768-36929768 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs953725230 Chr.22: 36929700-36929700 on Build GRCh38
rs770358540 Chr.22: 36929788-36929788 on Build GRCh38
rs61737702 Chr.22: 36929642-36929642 on Build GRCh38
rs746992696 Chr.22: 36929570-36929570 on Build GRCh38
rs539880950 Chr.22: 36929732-36929732 on Build GRCh38
rs939255243 Chr.22: 36929745-36929745 on Build GRCh38
rs753738312 Chr.22: 36929763-36929763 on Build GRCh38
rs751578225 Chr.22: 36929721-36929721 on Build GRCh38
rs367633751 Chr.22: 36929665-36929665 on Build GRCh38
rs779664566 Chr.22: 36929740-36929740 on Build GRCh38
rs113156338 Chr.22: 36929661-36929661 on Build GRCh38
rs754189724 Chr.22: 36929621-36929621 on Build GRCh38
rs757383286 Chr.22: 36929727-36929727 on Build GRCh38
rs563870106 Chr.22: 36929797-36929797 on Build GRCh38
rs139583249 Chr.22: 36929701-36929701 on Build GRCh38
rs771775634 Chr.22: 36929783-36929783 on Build GRCh38
rs775059195 Chr.22: 36929602-36929602 on Build GRCh38
rs748941609 Chr.22: 36929676-36929676 on Build GRCh38
rs763038738 Chr.22: 36929713-36929713 on Build GRCh38
rs764333642 Chr.22: 36929714-36929714 on Build GRCh38
rs866537612 Chr.22: 36929699-36929699 on Build GRCh38
rs768091118 Chr.22: 36929610-36929610 on Build GRCh38
rs372395367 Chr.22: 36929751-36929751 on Build GRCh38
rs762515992 Chr.22: 36929603-36929603 on Build GRCh38
rs529939805 Chr.22: 36929571-36929571 on Build GRCh38
rs370120556 Chr.22: 36929673-36929673 on Build GRCh38
rs747650265 Chr.22: 36929767-36929767 on Build GRCh38
rs966332244 Chr.22: 36929672-36929672 on Build GRCh38
rs761414020 Chr.22: 36929685-36929685 on Build GRCh38
rs780918629 Chr.22: 36929572-36929572 on Build GRCh38
rs145546133 Chr.22: 36929762-36929762 on Build GRCh38
rs140679574 Chr.22: 36929662-36929662 on Build GRCh38
rs879440876 Chr.22: 36929790-36929791 on Build GRCh38
rs771509643 Chr.22: 36929686-36929686 on Build GRCh38
rs781118627 Chr.22: 36929666-36929666 on Build GRCh38
rs893148604 Chr.22: 36929716-36929716 on Build GRCh38
rs540559131 Chr.22: 36929598-36929598 on Build GRCh38
rs367927682 Chr.22: 36929702-36929702 on Build GRCh38
rs376547021 Chr.22: 36929612-36929612 on Build GRCh38
rs180766723 Chr.22: 36929776-36929776 on Build GRCh38
rs150587930 Chr.22: 36929684-36929684 on Build GRCh38
rs777167803 Chr.22: 36929784-36929784 on Build GRCh38
rs777336576 Chr.22: 36929690-36929690 on Build GRCh38
rs762165060 Chr.22: 36929584-36929614 on Build GRCh38
rs766424551 Chr.22: 36929675-36929675 on Build GRCh38
rs765484350 Chr.22: 36929641-36929641 on Build GRCh38
rs755915142 Chr.22: 36929737-36929737 on Build GRCh38
rs773833290 Chr.22: 36929611-36929611 on Build GRCh38
rs745658011 Chr.22: 36929667-36929667 on Build GRCh38
rs16846 Chr.22: 36929791-36929791 on Build GRCh38
rs149714683 Chr.22: 36929733-36929733 on Build GRCh38
rs374827848 Chr.22: 36929683-36929683 on Build GRCh38
rs745663251 Chr.22: 36929591-36929591 on Build GRCh38
rs759942643 Chr.22: 36929693-36929693 on Build GRCh38
rs902433304 Chr.22: 36929590-36929590 on Build GRCh38
rs767022834 Chr.22: 36929619-36929619 on Build GRCh38
rs112325735 Chr.22: 36929648-36929648 on Build GRCh38
rs746415173 Chr.22: 36929787-36929787 on Build GRCh38
rs749137759 Chr.22: 36929750-36929750 on Build GRCh38
rs759831547 Chr.22: 36929632-36929632 on Build GRCh38

View Full Product Details