Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24303671-24303934 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOP9
Entrez Gene ID
161424
Gene Name
NOP9 nucleolar protein
Gene Aliases
C14orf21
UniGene
Hs.708040
Gene Chromosome Location
Chr.14: 24299854-24309126 on Build GRCh38
Transcripts
NM_001286367.1 NM_174913.2 XM_005267385.1 XM_011536526.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6317261 c.1313G>T p.R438L Chr.14: 24303760-24303760 on Build GRCh38
5504032 c.1310delC p.R438fs*12 Chr.14: 24303757-24303757 on Build GRCh38
4751370 c.1385A>G p.E462G Chr.14: 24303832-24303832 on Build GRCh38
3700847 c.1308A>G p.S436S Chr.14: 24303755-24303755 on Build GRCh38
698080 c.1335C>G p.L445L Chr.14: 24303782-24303782 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201048704 Chr.14: 24303882-24303882 on Build GRCh38
rs756810879 Chr.14: 24303892-24303892 on Build GRCh38
rs748875365 Chr.14: 24303698-24303698 on Build GRCh38
rs770331343 Chr.14: 24303701-24303701 on Build GRCh38
rs150879895 Chr.14: 24303823-24303823 on Build GRCh38
rs746396256 Chr.14: 24303807-24303807 on Build GRCh38
rs750485260 Chr.14: 24303851-24303851 on Build GRCh38
rs116960868 Chr.14: 24303759-24303759 on Build GRCh38
rs962782531 Chr.14: 24303788-24303788 on Build GRCh38
rs748996225 Chr.14: 24303797-24303797 on Build GRCh38
rs1025758127 Chr.14: 24303832-24303832 on Build GRCh38
rs777536803 Chr.14: 24303782-24303782 on Build GRCh38
rs772527305 Chr.14: 24303814-24303814 on Build GRCh38
rs765375976 Chr.14: 24303839-24303839 on Build GRCh38
rs776979755 Chr.14: 24303828-24303828 on Build GRCh38
rs372860523 Chr.14: 24303694-24303694 on Build GRCh38
rs775945837 Chr.14: 24303815-24303815 on Build GRCh38
rs1034642590 Chr.14: 24303773-24303773 on Build GRCh38
rs747993449 Chr.14: 24303765-24303765 on Build GRCh38
rs4982895 Chr.14: 24303716-24303716 on Build GRCh38
rs34064567 Chr.14: 24303732-24303732 on Build GRCh38
rs886373422 Chr.14: 24303744-24303744 on Build GRCh38
rs761076943 Chr.14: 24303817-24303817 on Build GRCh38
rs774987469 Chr.14: 24303704-24303704 on Build GRCh38
rs778425557 Chr.14: 24303908-24303908 on Build GRCh38
rs764417992 Chr.14: 24303718-24303718 on Build GRCh38
rs767881421 Chr.14: 24303713-24303713 on Build GRCh38
rs767564931 Chr.14: 24303849-24303850 on Build GRCh38
rs772705522 Chr.14: 24303697-24303697 on Build GRCh38
rs537549572 Chr.14: 24303726-24303726 on Build GRCh38
rs760919390 Chr.14: 24303717-24303717 on Build GRCh38
rs184642630 Chr.14: 24303766-24303766 on Build GRCh38
rs753540441 Chr.14: 24303884-24303884 on Build GRCh38
rs377676583 Chr.14: 24303869-24303869 on Build GRCh38
rs767607524 Chr.14: 24303864-24303864 on Build GRCh38
rs781159717 Chr.14: 24303764-24303764 on Build GRCh38
rs774471508 Chr.14: 24303824-24303826 on Build GRCh38
rs201790677 Chr.14: 24303793-24303793 on Build GRCh38
rs766205100 Chr.14: 24303826-24303827 on Build GRCh38
rs757417500 Chr.14: 24303738-24303738 on Build GRCh38
rs756000990 Chr.14: 24303876-24303876 on Build GRCh38
rs951482358 Chr.14: 24303842-24303842 on Build GRCh38
rs748884588 Chr.14: 24303789-24303789 on Build GRCh38
rs373890909 Chr.14: 24303866-24303866 on Build GRCh38
rs1001042178 Chr.14: 24303871-24303871 on Build GRCh38
rs765230403 Chr.14: 24303740-24303740 on Build GRCh38
rs759967138 Chr.14: 24303711-24303711 on Build GRCh38
rs763091974 Chr.14: 24303863-24303863 on Build GRCh38
rs553174329 Chr.14: 24303830-24303830 on Build GRCh38
rs769528552 Chr.14: 24303696-24303696 on Build GRCh38
rs1031146141 Chr.14: 24303911-24303911 on Build GRCh38
rs370914104 Chr.14: 24303904-24303904 on Build GRCh38

View Full Product Details