Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 127051912-127052137 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf63
Entrez Gene ID
401207
Gene Name
chromosome 5 open reading frame 63
Gene Aliases
YDR286C
UniGene
Hs.49573
Gene Chromosome Location
Chr.5: 127040752-127073539 on Build GRCh38
Transcripts
NM_001164478.1 NM_001164479.1 XM_017009464.1 XM_017009465.1

SNP Detail

SNP ID Chromosome Location
rs971661791 Chr.5: 127051959-127051959 on Build GRCh38
rs769539525 Chr.5: 127051976-127051976 on Build GRCh38
rs2240800 Chr.5: 127052002-127052002 on Build GRCh38
rs752441310 Chr.5: 127051984-127051984 on Build GRCh38
rs962925094 Chr.5: 127051987-127051987 on Build GRCh38
rs535174164 Chr.5: 127052034-127052034 on Build GRCh38
rs549242579 Chr.5: 127052052-127052052 on Build GRCh38
rs1020894411 Chr.5: 127052086-127052086 on Build GRCh38
rs777625284 Chr.5: 127051989-127051989 on Build GRCh38
rs73339251 Chr.5: 127052042-127052042 on Build GRCh38
rs770416990 Chr.5: 127051996-127051996 on Build GRCh38
rs929811668 Chr.5: 127051935-127051935 on Build GRCh38
rs945246373 Chr.5: 127052055-127052058 on Build GRCh38
rs755197576 Chr.5: 127051938-127051938 on Build GRCh38
rs1042314552 Chr.5: 127052071-127052071 on Build GRCh38
rs185821942 Chr.5: 127052005-127052005 on Build GRCh38
rs35778264 Chr.5: 127052015-127052016 on Build GRCh38
rs1004472127 Chr.5: 127051968-127051968 on Build GRCh38
rs1010390178 Chr.5: 127052081-127052081 on Build GRCh38
rs78618698 Chr.5: 127051990-127051990 on Build GRCh38
rs564514499 Chr.5: 127051985-127051985 on Build GRCh38
rs541039963 Chr.5: 127051939-127051939 on Build GRCh38
rs1047395433 Chr.5: 127051937-127051937 on Build GRCh38
rs954853255 Chr.5: 127052054-127052054 on Build GRCh38

View Full Product Details